Skip to Content
Merck
All Photos(1)

Documents

DFF100

Sigma-Aldrich

DEAE–Sepharose

Fast Flow

Synonym(s):

Diethylaminoethyl–Sepharose

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
47101511
NACRES:
NA.56

form

suspension

technique(s)

affinity chromatography: suitable

matrix

6% cross-linked agarose

bead size

45-165 μm (wet)

pore size

~4,000,000 Da exclusion limit

pH

2—12

capacity

110-160 μeq/mL binding capacity(gel volume)

Looking for similar products? Visit Product Comparison Guide

General description

DFF100-500Ml′s update product number is GE17-0709-01

Application

DEAE-Sepharose has been used in:
  • anion exchange chromatography
  • to screen polyisoprenyl phosphate phosphatase activity
  • to purify isoinhibitors
  • for the purification of human immunodeficiency virus (HIV)
  • glycoprotein envelope (gp140 env)

DEAE-Sepharose is used in affinity chromatography, protein chromatography and ion exchange chromatography. DEAE-Sepharose has been used to study pathogenesis of human disease and to develop a new assay for detecting the toxins of pathogenic strains of Clostridium difficile.

Biochem/physiol Actions

Diethylaminoethyl-sepharose (DEAE-sepharose) is a strong anion exchange column, where DEAE covalently binds to sepharose.

Legal Information

Sepharose is a trademark of Cytiva

replaced by

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

100.4 - 109.4 °F

Flash Point(C)

38 - 43 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G L Yewey et al.
Biochemical and biophysical research communications, 148(3), 1520-1526 (1987-11-13)
Bovine heart cytochrome c oxidase, depleted of polypeptide subunits by alkaline detergent treatment, was characterized with respect to metal content, optical spectral properties, and oxidase activity. Treatment with 1.0% Triton X-100 at pH 9.5 followed by anion-exchange chromatography caused removal
Jessie Fernandez et al.
mBio, 12(1) (2021-02-11)
Rice blast disease caused by Magnaporthe oryzae is a devastating disease of cultivated rice worldwide. Infections by this fungus lead to a significant reduction in rice yields and threats to food security. To gain better insight into growth and cell
Rapid high-level production of functional HIV broadly neutralizing monoclonal antibodies in transient plant expression systems
Rosenberg Y, et al.
Testing, 8(3), e58724-e58724 (2013)
Effective Purification Procedure of <I>Aspergillus oryzae</I> α−Amylase from Solid State Fermentation Cultures Including Concanavalin-A Sepharose
Terebiznik, M.R., et al.
Journal of Food Biochemistry, 19(5), 341-354 (1995)
Growth cone collapse and neurite retraction from cultured <I>Helisoma</I> neurons in response to antibody Fab fragments against an extracellular matrix protein.
Miller, J.D., et al.
Dev. Brain Res., 79(2), 203-218 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service