Skip to Content
Merck
All Photos(1)

Key Documents

GE17-0709-10

DEAE Sepharose Fast Flow

Cytiva 17-0709-10, pack of 25 mL

Synonym(s):

Anion exchange purification

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.56

ligand

diethylaminoethyl

description

Ion Exchanger Type (value)

packaging

pack of 25 mL

manufacturer/tradename

Cytiva 17-0709-10

matrix

6% cross-linked agarose

particle size

45-165 μm

average diameter

90 μm

cleaning in place

2-14

working range

2-12

suitability

suitable for bioprocess medium

Related Categories

General description

DEAE Sepharose Fast FLow is a weak anion exchanger based on the well established Sepharose Fast FLow ion exchange platform, extensively used for preparative protein separations in both research and industrial applications.

Application

DEAE Sepharose Fast FLow is part of the Sepharose Fast FLow ion exchange platform, which has been the industrial standard for ion exchange chromatography during recent decades. It is composed of crosslinked 6% Agarose beads, with diethylaminoethyl (DEAE) weak anion exchange groups. DEAE Sepharose Fast FLow has high chemical stability, allowing well proven cleaning-in-place (CIP) and sanitization protocols. Scale-up with DEAE Sepharose Fast FLow is straight forward and the medium is available in a range of pre-packed process development tools.

As member of the BioProcess media range, DEAE Sepharose Fast FLow meets industrial demands with security of supply and comprehensive technical and regulatory support.

Features and Benefits

  • Well-proven weak anion exchanger developed for industrial downstream processes
  • Used extensively for capture and intermediate purification of a wide range of approved biopharmaceuticals
  • The industry standard for ion exchange chromatography during recent decades
  • High chemical stability allows for well proven CIP and sanitization protocols
  • The hydrophilic nature of the base matrix ensures Low levels of non-specific binding leading to Low levels of host cell-derived impurities in the elution pool.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service