Skip to Content
Merck

IP50

Protein G Immunoprecipitation Kit

sufficient for 50 assays

Synonym(s):

Immunoprecipitation kit

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

NACRES:
NA.32
UNSPSC Code:
12352203
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

usage

sufficient for 50 assays

Quality Level

technique(s)

immunoprecipitation (IP): suitable

storage temp.

2-8°C

Application

The kit is designed to allow maximal recovery of immunoprecipitates. It provides all the necessary reagents to perform immunoprecipitation from cell extracts of any protein to which a suitable antibody is available. Based on protein G, the kit binds to most commonly used antibodies. In addition, spin columns are provided to enable quick washes without the loss of protein G resin and thus protein yield is maximized.

Features and Benefits

  • Minimal loss of antigen-antibody bound beads during washing.
  • Minimal or no non-specific signals by increasing the stringency of the washing step.

Preparation Note

When preparing reagents, use ultrapure water (17M -cm)

Kit Components Also Available Separately

Product No.
Description
SDS

  • S65465 M Sodium chloride solution 15 mLSDS

  • P3296Protein G Agarose 2 mLSDS

  • 7173610% Sodium dodecyl sulfate solution 1 mLSDS

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patricia Workman et al.
Journal of bacteriology, 194(13), 3512-3521 (2012-05-01)
The BamA protein of Escherichia coli plays a central role in the assembly of β-barrel outer membrane proteins (OMPs). The C-terminal domain of BamA folds into an integral outer membrane β-barrel, and the N terminus forms a periplasmic polypeptide transport-associated
Attenuated PGI2 synthesis in obese Zucker rats
Hodnett,B et al.
American Journal of Physiology. Regulatory, Integrative and Comparative Physiology, 296(3), R715-R721 (2010)
Hubert Arokium et al.
The Journal of biological chemistry, 282(48), 35104-35112 (2007-10-04)
During apoptosis, the pro-apoptotic protein Bax relocalizes from the cytosol to the mitochondrial outer membrane. This relocalization is associated to major conformational changes, namely at the N- and C-terminal ends of the protein. Substitution of residues located at critical positions
F Gautier et al.
Molecular and cellular biology, 31(4), 832-844 (2010-12-22)
Bcl-2 homologues (such as Bcl-x(L)) promote survival in part through sequestration of "activator" BH3-only proteins (such as Puma), preventing them from directly activating Bax. It is thus assumed that inhibition of interactions between activators and Bcl-x(L) is a prerequisite for
R Meller et al.
Cell death and differentiation, 10(5), 539-547 (2003-05-03)
Seizure-induced neuronal death may involve engagement of the BCL-2 family of apoptosis-regulating proteins. In the present study we examined the activation of proapoptotic BAD in cultured hippocampal neurons following seizures induced by removal of chronic glutamatergic transmission blockade. Kynurenic acid

Global Trade Item Number

SKUGTIN
IP50-1KT04061833866092

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service