Skip to Content
Merck
All Photos(2)

Documents

GE17-0405-01

HiTrap® Protein G High Performance

Cytiva 17-0405-01, pack of 1 × 5 mL

Synonym(s):

Protein G affinity column, Protein purification column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151816
NACRES:
NA.56

ligand

recombinant protein G lacking albumin-binding region

packaging

pack of 1 × 5 mL

manufacturer/tradename

Cytiva 17-0405-01

storage condition

(20% Ehtanol)
(20% Ethanol)

parameter

flow rate
42 psi

bed size

16 mm × 25 mm

bed volume

5 mL

column I.D.

16 mm

matrix

highly cross-linked 6% agarose

avg. part. size

34 μm

cleaning

2-9

working range

3-9

capacity

>1 mg binding capacity (lgG/column)(The binding capacity has been determined with human polyclonal IgG.)
125 mg binding capacity(human IgG/column)

storage temp.

2-8°C

General description

HiTrap® Protein G HP columns are prepacked with Protein G Sepharose High Performance for fast, easy separations of IgG from different species and subclasses.
pH below 3 is sometimes required to elute strongly bound IgG species. However, protein ligands may hydrolyze at very low pH.

Features and Benefits

  • High binding capacity of 25 mg IgG/mL medium and fast binding kinetics.
  • 34 μm bead size for high performance, high-resolution purifications.
  • Binds broad range of different species and subclasses of IgG.
  • Simple and proven method giving reproducible results.
  • Designed for use with syringe, peristalic pump, or chromatography system.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

HiTrap is a registered trademark of Cytiva
Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wioleta M Zelek et al.
Immunology, 161(2), 103-113 (2020-06-20)
The implication of complement in multiple diseases over the last 20 years has fuelled interest in developing anti-complement drugs. To date, the focus has been on C5; blocking cleavage of C5 prevents formation of two pro-inflammatory activities, C5a anaphylatoxin and membrane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service