Skip to Content
Merck
All Photos(1)

Key Documents

D9663

Sigma-Aldrich

Donkey serum

Synonym(s):

Serum

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
biological source:
donkey
sterility:
non-sterile
shipped in:
dry ice

biological source

donkey

Quality Level

sterility

non-sterile

contains

preservative

application(s)

research pathology

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Donkey serum is pooled from a normal donor herd. In immunoassays, normal donkey serum can serve as a blocking agent and a control. It is used as a blocking agent in various studies for immunohistochemistry and immunocytochemistry, ranging from 5% to 20%.

Application

Donkey serum has been used:
  • as a component in blocking buffer for immunofluorescence
  • in phosphate buffer saline (PBS) as a blocking agent
  • as a blocking agent in frozen immunocytochemistry and immunohistochemistry
  • to prevent nonspecific binding in parvalbumin immunohistochemistry

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

related product

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marianne O Larsen et al.
American journal of physiology. Endocrinology and metabolism, 282(6), E1342-E1351 (2002-05-15)
Nonrodent models of diabetes are needed for practical and physiological reasons. Induction of mild insulin-deficient diabetes was investigated in male Göttingen minipigs by use of streptozotocin (STZ) alone (75, 100, and 125 mg/kg) or 125 mg/kg combined with pretreatment with
Omid Ahmadi et al.
Journal of gastroenterology and hepatology, 25(2), 277-285 (2009-10-02)
Interstitial cells of Cajal (ICC) are distributed with smooth muscle throughout the gastrointestinal tract and are involved in regulating motility. ICC were recently discovered in the wall of the human gallbladder. This study sought to determine whether ICC are present
Yasmin Fardghassemi et al.
Disease models & mechanisms, 10(12), 1465-1480 (2017-10-25)
Polyglutamine expansion diseases are a group of hereditary neurodegenerative disorders that develop when a CAG repeat in the causative genes is unstably expanded above a certain threshold. The expansion of trinucleotide CAG repeats causes hereditary adult-onset neurodegenerative disorders, such as
Helen F Irving-Rodgers et al.
Biology of reproduction, 69(6), 2022-2028 (2003-08-22)
Bovine ovarian antral follicles exhibit either one or the other of two patterns of granulosa cell death in atresia. Death can commence either from the antrum and progress toward the basal lamina (antral atresia) or the converse (basal atresia). In
H F Irving-Rodgers et al.
Molecular reproduction and development, 73(10), 1292-1302 (2006-07-26)
Using immunohistochemistry and RNA analyses we examined the fate of components of a newly identified matrix that develops between granulosa cells (focimatrix, abbreviated from focal intraepithelial matrix) and of the follicular basal lamina in ovulating bovine ovarian follicles. Pre- and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service