All Photos(1)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.46
biological source:
goat
sterility:
non-sterile
shipped in:
dry ice
Recommended Products
biological source
goat
Quality Level
sterility
non-sterile
contains
preservative
application(s)
research pathology
shipped in
dry ice
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
General description
Goat serum is pooled from a normal donor herd.
Application
Goat serum has been used in:
- immunohistochemistry
- immunocytochemistry
- double immunofluorescence
- enzyme immunoassays (EIAs)
Normal goat serum may be used as a blocking agent or negative control in immunoassays. For immunohistochemistry and immunocytochemistry, goat serum was used between 1%-20% as blocking agent in various studies.
Biochem/physiol Actions
Goat serum can be effectively used for routine culture of fish cells.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
related product
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
John R Phillips et al.
Investigative ophthalmology & visual science, 45(3), 758-763 (2004-02-27)
To investigate the change in axial eye dimensions resulting from stretching the sclera by acute elevation of intraocular pressure (IOP). IOP was increased to 100 mm Hg for 1 hour through an intravitreal cannula, while ocular component dimensions were monitored
Pressure-induced changes in axial eye length of chick and tree shrew: significance of myofibroblasts in the sclera
Phillips J R and McBrien N A
Investigative Ophthalmology & Visual Science, 45(3), 758-763 (2004)
T O Schneider et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 17(2), 192-199 (1999-04-30)
The objective of this study was to investigate the presence of a contractile actin isoform, alpha-smooth muscle actin, in annulus fibrosus cells in situ and in two and three-dimensional cultures. Annulus fibrosus cells were isolated from healthy adult dogs, serial
Tendon cell contraction of collagen-GAG matrices in vitro: effect of cross-linking
Torres DS, et al.
Biomaterials, 21(15), 1607-1619 (2000)
Performance of a time-resolved fluorescence immunoassay for measuring varicella-zoster virus immunoglobulin G levels in adults and comparison with commercial enzyme immunoassays and Merck glycoprotein enzyme immunoassay
Maple P A C, et al.
Clinical and Vaccine Immunology : CVI, 13(2), 214-218 (2006)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service