Skip to Content
Merck
All Photos(1)

Key Documents

G6767

Sigma-Aldrich

Goat Serum Donor Herd

USA origin, sterile-filtered, suitable for cell culture

Synonym(s):

caprine sera, caprine serum, goat sera

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

biological source

goat

Quality Level

sterility

sterile-filtered

composition

hemoglobin, ≤30 mg/dL

origin

USA origin

technique(s)

cell culture | mammalian: suitable

impurities

≤20 EU/mL endotoxin

shipped in

dry ice

storage temp.

−20°C

General description

Goat serum is a suitable alternative to fetal bovine serum in a wide range of cell culture applications such as the isolation and identification of viruses produced in a variety of primary cell lines.

Application

Goat Serum Donor Herd has been used as a blocking solution in:
  • immunocytostaining
  • immunofluorescence staining
  • immunohistochemistry

Preparation Note

Collected from a controlled donor herd.

Analysis Note

Endotoxin tested

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruoxi Zhang et al.
Virology, 485, 414-421 (2015-09-08)
To subvert host antiviral immune responses, many viruses have evolved countermeasures to inhibit IFN signaling pathway. Porcine bocavirus (PBoV), a newly identified porcine parvovirus, has received attention because it shows clinically high co-infection prevalence with other pathogens in post-weaning multisystemic
Meghan E Breitbach et al.
PLoS pathogens, 15(8), e1007766-e1007766 (2019-08-02)
Zika virus (ZIKV) and dengue virus (DENV) are genetically and antigenically related flaviviruses that now co-circulate in much of the tropical and subtropical world. The rapid emergence of ZIKV in the Americas in 2015 and 2016, and its recent associations
Ruoxi Zhang et al.
Virus genes, 52(6), 797-805 (2016-11-01)
Type I interferon (IFN) and the IFN-induced cellular antiviral responses are the primary defense mechanisms against viral infection; however, viruses always evolve various mechanisms to antagonize this host's IFN responses. Porcine bocavirus (PBoV) is a newly identified porcine parvovirus. In
Smita Paranjape
Indian journal of experimental biology, 42(1), 26-35 (2004-07-28)
Serum is frequently added to the defined basal medium as a source of certain nutritional and macromolecular growth factors essential for cell growth. Although a number of synthetic media have been prepared serum continues to be used in cell culture
Runxia H Wen et al.
Autophagy, 15(11), 1970-1989 (2019-04-13)
We previously reported autophagic structures in rod photoreceptors expressing a misfolding RHO (rhodopsin) mutant (RHOP23H), suggesting that autophagy may play a role in degrading the mutant RHO and/or be involved in photoreceptor cell death. To further examine autophagy in normal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service