Skip to Content
Merck
All Photos(5)

Documents

D8001

Sigma-Aldrich

3,3′-Diaminobenzidine

97% (HPLC)

Synonym(s):

3,3′,4,4′-Biphenyltetramine, 3,3′,4,4′-Tetraaminobiphenyl, DAB

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH2)2C6H3C6H3(NH2)2
CAS Number:
Molecular Weight:
214.27
Beilstein:
1212988
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

Assay

97% (HPLC)

form

powder

mp

175-177 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

NC1=C(N)C=CC(C2=CC(N)=C(N)C=C2)=C1

InChI

1S/C12H14N4/c13-9-3-1-7(5-11(9)15)8-2-4-10(14)12(16)6-8/h1-6H,13-16H2

InChI key

HSTOKWSFWGCZMH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,3′-Diaminobenzidine has been used as a chromogen for the visualization in immunohistochemistry of chondrosarcoma sections, mammary carcinomas and brain tissue sections.
Peroxidase substrate; reagent for spectrophotometric determination of selenium.

Biochem/physiol Actions

DAB (3,3′-Diaminobenzidine) is utilized in many applications for the visualization of peroxidase activity. In the peroxidase reaction, DAB serves as a hydrogen donor in the presence of peroxide. The oxidized DAB forms an insoluble brown end-product for use in the immunohistological and immunoblotting staining procedures.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A colorimetric method for measurement of the (peroxidase-mediated) oxidation of 3,3'-diaminobenzidine.
Fahimi HD and Herzog V
The Journal of Histochemistry and Cytochemistry, 21, 499-502 (1973)
Pathogenesis of a Thai strain of white spot syndrome virus (WSSV) in juvenile, specific pathogen-free Litopenaeus vannamei.
Escobedo-Bonilla CM, et al.
Diseases of Aquatic Organisms, 74, 85-94 (2007)
Detection of T lymphocytes in intestine of broiler chicks treated with Lactobacillus spp. and challenged with Salmonella enterica serovar Enteritidis.
Noujaim JC, et al.
Poultry Science, 87, 927-933 (2008)
Metastatic canine mammary carcinomas can be identified by a gene expression profile that partly overlaps with human breast cancer profiles
Klopfleisch R, et al.
BMC Cancer, 10(1), 618-618 (2010)
Motor skill training promotes sensorimotor recovery and increases microtubule-associated protein-2 (MAP-2) immunoreactivity in the motor cortex after intracerebral hemorrhage in the rat
Santos MV, et al.
ISRN neurology, 2013 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service