Skip to Content
Merck
All Photos(1)

Documents

D4418

Sigma-Aldrich

3,3′-Diaminobenzidine

SIGMAFAST, chromogenic, tablet

Synonym(s):

DAB, DAB tablets, Immunohistology substrate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

product name

SIGMAFAST 3,3′-Diaminobenzidine tablets, tablet, To prepare 15 mL

form

tablet

solubility

water: 1 tablet/5 mL

storage temp.

−20°C

SMILES string

Nc1ccc(cc1N)-c2ccc(N)c(N)c2

InChI

1S/C12H14N4/c13-9-3-1-7(5-11(9)15)8-2-4-10(14)12(16)6-8/h1-6H,13-16H2

InChI key

HSTOKWSFWGCZMH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Developed for use in immunohistology and immunoblotting as a precipitating substrate for the localization of peroxidase activity. DAB is the immunohistology substrate of choice as it produces an intense brown-black stain which is resistant to alcohol. Slides stained with DAB may be dehydrated, mounted in resinous media, and stored for future reference. SIGMAFAST DAB Tablets require no additional buffers or steps to prepare an active substrate solution. Tablets are individually packaged in foil packets

Reconstitution

Each tablet set dissolved in 15 mL deionized water yields a ready-to-use buffered solution containing DAB and urea hydrogen peroxide.

Legal Information

SIGMAFAST is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Muta. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erin N Scully et al.
Behavioural brain research, 331, 151-158 (2017-05-17)
Zebra finches (Taeniopygia guttata) are sexually dimorphic songbirds, not only in appearance but also in vocal production: while males produce both calls and songs, females only produce calls. This dimorphism provides a means to contrast the auditory perception of vocalizations
Huiming Yang et al.
Nature communications, 11(1), 2582-2582 (2020-05-24)
Polyglutamine expansion in proteins can cause selective neurodegeneration, although the mechanisms are not fully understood. In Huntington's disease (HD), proteolytic processing generates toxic N-terminal huntingtin (HTT) fragments that preferentially kill striatal neurons. Here, using CRISPR/Cas9 to truncate full-length mutant HTT
Monika Verma et al.
Toxicon : official journal of the International Society on Toxinology, 139, 58-65 (2017-09-19)
Anthrax associated causalities in humans and animals are implicated mainly due to the action of two exotoxins that are secreted by the bacterium Bacillus antharcis during the infection. These exotoxins comprise of three protein components namely protective antigen (PA), lethal
Laura Riva et al.
Pathogens (Basel, Switzerland), 10(2) (2021-02-10)
The hepatitis C virus (HCV) life cycle is a tightly regulated process, during which structural and non-structural proteins cooperate. However, the interplay between HCV proteins during genomic RNA replication and progeny virion assembly is not completely understood. Here, we studied
Ya-Chao Wang et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 37(3), 980-993 (2016-05-14)
The NMDA antagonist memantine preferentially inhibits extrasynaptic NMDA receptors, which are overactivated upon stroke and thought to disturb neuroplasticity. We hypothesized that memantine enhances post-ischemic neurological recovery, brain remodeling, and plasticity. C57BL6/j mice were exposed to intraluminal middle cerebral artery

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service