Skip to Content
Merck
All Photos(1)

Key Documents

D6815

Sigma-Aldrich

3,3′-Diaminobenzidine (DAB) Enhanced Liquid Substrate System tetrahydrochloride

peroxidase substrate, liquid, suitable for membrane applications

Synonym(s):

DAB membrane substrate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Product Name

3,3′-Diaminobenzidine (DAB) Enhanced Liquid Substrate System tetrahydrochloride, for Membrane Applications

Quality Level

form

liquid

shipped in

wet ice

storage temp.

2-8°C

SMILES string

Cl.Cl.Cl.Cl.Nc1ccc(cc1N)-c2ccc(N)c(N)c2

InChI

1S/C12H14N4.4ClH/c13-9-3-1-7(5-11(9)15)8-2-4-10(14)12(16)6-8;;;;/h1-6H,13-16H2;4*1H

InChI key

KJDSORYAHBAGPP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,3′-Diaminobenzidine (DAB) Enhanced Liquid Substrate System tetrahydrochloride has been used as a substrate for peroxidase coupled secondary antibody.
  • in paraffin heart sections of mice for immunohistochemical and immunofluorescent staining
  • in immunotransfer of salivary protein of laboratory Lutzomyia ovallesi
  • in immunoperoxidase staining of lung tissue
The DAB (3,3′-Diaminobenzidine tetrahydrochloride) Two-component Enhanced Liquid Substrate System is a precipitating substrate system for peroxidase. DAB Two-component Liquid Substrate System produces a brown precipitate for use in immunohistological and immunoblotting staining procedures.

Biochem/physiol Actions

3,3′-Diaminobenzidine (DAB) is a chromogenic substrate for peroxidase enzyme. It is converted to an insoluble precipitate after oxidation by peroxidase. This feature is exploited for visualization in histochemical (HC) and immunohistochemical (IHC) staining.

Packaging

Set consists of 4 x 100 ml Solution A and 15 ml Solution B.

Caution

Stable for up to 18 mo. when stored at 2-8 °C in the original container.

Quantity

Each DAB Two-component Liquid Substrate System will provide 500 mL of the DAB substrate solution.

Kit Components Also Available Separately

Product No.
Description
SDS

  • 3, 3′-Diaminobenzidine enhanced liquid substrate system 4 x 100

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Muta. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Władysław Polcyn
Acta biochimica Polonica, 55(4), 761-766 (2008-11-19)
Previously, we showed that anaerobic induction of respiratory nitrate reductase (NR) activity in Bradyrhizobium sp. (Lupinus) USDA 3045 is strongly enhanced by nitrate or nitrite through de novo synthesis. Here, multiple NR-active soluble forms, ranging from 75 kDa to 190
Preclinical efficacy studies of influenza A haemagglutinin precursor cleavage loop peptides as a potential vaccine
Miller DS, et al.
The Journal of General Virology, 92, 1152-1161 (2011)
Acute erythropoietin cardioprotection is mediated by endothelial response
Teng R, et al.
Basic Research in Cardiology, 106, 343-354 (2011)
Preclinical efficacy studies of influenza A haemagglutinin precursor cleavage loop peptides as a potential vaccine
Miller DS, et al.
The Journal of General Virology, 92(5), 1152-1161 (2011)
Nanette Mittereder et al.
Journal of immunology (Baltimore, Md. : 1950), 197(2), 491-503 (2016-06-15)
ICOS, a member of the CD28 family, represents a key molecule that regulates adaptive responses to foreign Ags. ICOS is prominently expressed on T follicular helper (TFH) cells, a specialized CD4(+) T cell subset that orchestrates B cell differentiation within

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service