Skip to Content
Merck
All Photos(1)

Key Documents

D0426

Sigma-Aldrich

DAB with Metal Enhancer

SIGMAFAST, peroxidase substrate, tablet

Synonym(s):

3,3′-Diaminobenzidine Peroxidase Substrate, DAB Peroxidase Substrate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.83

Product Name

SIGMAFAST DAB with Metal Enhancer, tablet

form

tablet

solubility

water: 1 tablet/5 mL

storage temp.

−20°C

General description

SIGMAFAST DAB with Metal Enhancer Tablets require no additional buffers or steps to prepare an active substrate solution for detection of peroxidase activity. Commom use is in immunohistology as a precipitating substrate.

Application

SIGMAFAST DAB with Metal Enhancer has been used in the visualization of lung tissue sections using immunohistochemistry, dorsal root ganglion (DRG) neurons and immunohistochemical staining for collagen in anterior cruciate ligament (ACL) cells.
The DAB reaction is enhanced by the addition of CoCl2 and produces a more intense stain which is resistant to alcohol.

Reconstitution

Each tablet set dissolved in 5 mL deionized water yields a ready-to-use buffered solution containing DAB with a metal enhancer.

Legal Information

SIGMAFAST is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Carc. 1B - Eye Dam. 1 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Somanshu Banerjee et al.
Journal of photochemistry and photobiology. B, Biology, 175, 254-268 (2017-09-20)
Birds time their daily and seasonal activities in synchronization with circadian and annual periodicities in the environment, which is mainly provided by changes in photoperiod/day length conditions. Photoperiod appears to act at the level of eye, pineal and encephalic/deep brain
M Breijo et al.
Journal of economic entomology, 110(3), 1390-1393 (2017-04-08)
The horn fly, Haematobia irritans (L.), is a blood-sucking livestock ectoparasite responsible for substantial livestock losses. In the present work, the potential use of recombinant hematobin (HTB), a horn fly salivary protein, as an antigen for cattle vaccination was investigated.
Saba Shahin et al.
Toxicological sciences : an official journal of the Society of Toxicology, 161(2), 349-374 (2017-10-27)
Microwave (MW) radiation induced oxidative stress reduces dendritic arborization, spine density and number of hippocampal pyramidal neurons and hence, impair learning and spatial memory through p53-dependent/independent apoptosis of hippocampal neuronal and nonneuronal cells. However, the mechanisms responsible for MW radiation
Keith Morris-Schaffer et al.
Particle and fibre toxicology, 16(1), 1-1 (2019-01-08)
Recent epidemiological studies indicate early-life exposure to pollution particulate is associated with adverse neurodevelopmental outcomes. The need is arising to evaluate the risks conferred by individual components and sources of air pollution to provide a framework for the regulation of
Developing rat lung has a sided pacemaker region for morphogenesis-related airway peristalsis
Jesudason EC, et al.
American Journal of Respiratory Cell and Molecular Biology, 32(2), 118-127 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service