Skip to Content
Merck
All Photos(1)

Key Documents

R1383

Sigma-Aldrich

RPMI-1640 Medium

With ʟ-glutamine, without glucose and sodium bicarbonate, powder, suitable for cell culture

Synonym(s):

Roswell Park Memorial Institute 1640 medium

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

Product Name

RPMI-1640 Medium, With L-glutamine, without glucose and sodium bicarbonate, powder, suitable for cell culture

form

powder

technique(s)

cell culture | mammalian: suitable

components

L-glutamine: yes
phenol red: yes
sodium pyruvate: no
HEPES: no
NaHCO3: no

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

RPMI 1640 Medium was developed at Roswell Park Memorial Institute in 1966 by Moore and his co-workers. A modification of McCoy′s 5A Medium, it was formulated to support lymphoblastoid cells in suspension culture, but it has since been shown to support a wide variety of cells that are anchorage-dependent. Originally intended to be used with a serum supplement, RPMI 1640 has been shown to support several cell lines in the absence of serum. It has also been widely used in fusion protocols and in the growth of hybrid cells. This medium is suitable for culturing human normal and neoplastic leukocytes.

Application

RPMI-1640 Medium has been used as a cell culture medium to culture human MCF-7 breast cancer cells and human castrate-resistant prostate cancer (CRPC) cell lines for various cell culture experiments. It has also been used to culture human islets to assess the oxidative stress response in primary human islets.

Quantity

Formulated to contain 8.4 grams of powder per liter of medium.

Reconstitution

Supplement with 2.0 g/L sodium bicarbonate.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Renée J H A Tillie et al.
Frontiers in cell and developmental biology, 9, 695684-695684 (2021-08-31)
The protein 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase-3 (PFKFB3) is a key stimulator of glycolytic flux. Systemic, partial PFKFB3 inhibition previously decreased total plaque burden and increased plaque stability. However, it is unclear which cell type conferred these positive effects. Myeloid cells play an important
Jiayi Yao et al.
Cancers, 12(8) (2020-08-09)
The acidic pH of the tumor microenvironment plays a critical role in driving cancer development toward a more aggressive phenotype, but the underlying mechanisms are unclear. To this end, phenotypic and genotypic changes induced by adaptation of cancer cells to
Qinxi Ma et al.
mSphere, 4(3) (2019-06-28)
Fatty acids have known antifungal effects and are used in over-the-counter topical treatments. Screening of a collection of gene knockouts in Candida albicans revealed that one strain, carrying a deletion of the transcription factor DAL81, is very susceptible to the
Mette Flinck et al.
Cell cycle (Georgetown, Tex.), 17(9), 1056-1067 (2018-06-14)
Precise acid-base homeostasis is essential for maintaining normal cell proliferation and growth. Conversely, dysregulated acid-base homeostasis, with increased acid extrusion and marked extracellular acidification, is an enabling feature of solid tumors, yet the mechanisms through which intra- and extracellular pH
Mark Y Jeng et al.
The Journal of experimental medicine, 215(1), 51-62 (2017-12-02)
The expansion of CD8+CD28- T cells, a population of terminally differentiated memory T cells, is one of the most consistent immunological changes in humans during aging. CD8+CD28- T cells are highly cytotoxic, and their frequency is linked to many age-related

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service