Skip to Content
Merck
All Photos(1)

Key Documents

D2429

Sigma-Aldrich

DMEM - low glucose

With sodium pyruvate, without ʟ-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable fore cell culture

Synonym(s):

DME, Dulbecco′s Modified Eagle′s Medium - high glucose, DMEM

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

Product Name

Dulbecco′s Modified Eagle′s Medium - low glucose, 10 ×, With 1000 mg/L glucose (1x), without L-glutamine, sodium bicarbonate, and folic acid, liquid, sterile-filtered, suitable for cell culture

sterility

sterile-filtered

form

liquid

concentration

10 ×

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

NaHCO3: no
sodium pyruvate: yes
phenol red: yes
glucose: low
L-glutamine: no
HEPES: no

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Dulbecco′s Modified Eagle′s Medium (DME) is a modification of basal medium Eagle (BME) that contains a 4-fold higher concentration of amino acids and vitamins, as well as additional supplementary components. The original DME formula, first reported for culturing embryonic mouse cells, contained 1,000 mg/L of glucose.

Application

Dulbecco′s Modified Eagle′s Medium-low glucose has been used:
  • during organoid embedding for neutralizing thawed collagen I
  • to culture melanoma cancer cell lines
  • to store the cancer tissue samples
  • as a differentiation culture medium for C2C12 cells

Reconstitution

Supplement with 3.7 g/L sodium bicarbonate, 0.584 g/L L-glutamine, 0.004 g/L folic acid, at 1×.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tejas Shah et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 84, 526-534 (2016-10-04)
Folic acid (FA) and iron are essential supplements during pregnancy. Similarly effects of vitamin B12 (B12) inadequacy and high folate and low B12 status, on pregnancy outcome are available. However there are no mandatory recommendations for B12. There are many
Yi Li et al.
Physiological genomics, 50(3), 158-168 (2018-01-18)
Folic acid supplements taken during pregnancy can prevent neural tube defects and other developmental abnormalities. Here, we explored the effects of folate supplementation on gene expression and DNA methylation during C2C12 differentiation. Based on the folic acid concentration, this study
Richard Schwab et al.
Assay and drug development technologies, 5(4), 541-550 (2007-09-05)
Determination of multidrug resistance (MDR) activity of tumor cells could provide important information for the personalized therapy of cancer patients. The functional calcein assay (MultiDrug Quant Assay, Solvo Biotechnology, Budaörs, Hungary) has been proven to be clinically valuable in hematological
Maria Guix et al.
Scientific reports, 6, 21701-21701 (2016-02-26)
Novel approaches to develop naturally-induced drug delivery in tumor environments in a deterministic and controlled manner have become of growing interest in recent years. Different polymeric-based microstructures and other biocompatible substances have been studied taking advantage of lactic acidosis phenomena
Kévin Gillois et al.
The Science of the total environment, 754, 142324-142324 (2020-12-02)
Colloidal silver products are sold for a wide range of disinfectant and health applications. This has increased the potential for human exposure to silver nanoparticles (AgNPs) and ions (Ag+), for which oral ingestion is considered to be a major route

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service