Skip to Content
Merck
All Photos(2)

Documents

C2139

Sigma-Aldrich

Collagenase from Clostridium histolyticum

suitable for release of rat epididymal adipocytes and hepatocytes (for methodology see Type II and Type IV), Type VIII, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid

Synonym(s):

Clostridiopeptidase A

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

Clostridium histolyticum

Quality Level

type

Type VIII

form

powder

specific activity

≥125 CDU/mg solid
0.5-5.0 FALGPA units/mg solid

mol wt

68-125 kDa

suitability

suitable for release of rat epididymal adipocytes and hepatocytes (for methodology see Type II and Type IV)

application(s)

diagnostic assay manufacturing

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Clostridium histolyticum is a pathogenic clostridium that produces crude collagenases. This is a mixture of enzymes containing collagenase, non-specific proteases and clostripain.

Application

Collagenase may be used:
  • for the preparation of arterial tissue for the study of advanced glycosylation end products (AGE)
  • for use along with other proteases for the disaggregation of human tumor, mouse kidney, human brain, lung epithelium and many other tissues.
  • in liver and kidney perfusion studies, digestion of pancreas, and isolation of nonparenchymal hepatocytes.
  • for the preparation of viable hepatocytes from rat liver and for the isolation of fat cells from rat adipose tissue

Biochem/physiol Actions

Collagenase is activated by four gram atom calcium per mole enzyme. It is inhibited by ethylene glycol-bis(β-aminoethyl ether) - N, N, N′,N′-tetraacetic acid, β-mercaptoethanol, glutathione, thioglycolic acid and 8-hydroxyquinoline. Collagenase enzymes and neutral protease plays an important role in the effective release of cells from tissue. Collagenase recognizes the sequence -R-Pro-8-X-Gly-Pro-R-, where X represents a neutral amino acid.
Collagenase is activated by four gram atom calcium per mole enzyme. It is inhibited by ethylene glycol-bis(beta-aminoethyl ether) - N, N, N′,N′-tetraacetic acid, beta-mercaptoethanol, glutathione, thioglycolic acid and 8-hydroxyquinoline.

Unit Definition

One collagen digestion unit (CDU) liberates peptides from collagen from bovine achilles tendon equivalent in ninhydrin color to 1.0 μmole of leucine in 5 hours at pH 7.4 at 37 °C in the presence of calcium ions. One FALGPA hydrolysis unit hydrolyzes 1.0 μmole of furylacryloyl-Leu-Gly-Pro-Ala per min at 25°C. One Neutral Protease unit hydrolyzes casein to produce color equivalent to 1.0 μmole of tyrosine per 5 hr at pH 7.5 at 37°C. One Clostripain Unit hydrolyzes 1.0 μmole of BAEE per min at pH 7.6 at 25°C in the presence of DTT.

Analysis Note

Also contains clostripain, nonspecific neutral protease and tryptic activities.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fleischer, S. and B. Fleischer
Methods in Enzymology, 192, 829-829 (1990)
Ce Tang et al.
Nature immunology, 19(7), 755-765 (2018-06-20)
The cytokines IL-17A and IL-17F have 50% amino-acid identity and bind the same receptor; however, their functional differences have remained obscure. Here we found that Il17f-/- mice resisted chemically induced colitis, but Il17a-/- mice did not, and that Il17f-/- CD45RBhiCD4+
Fleischer, S. and B. Fleischer
Methods in Enzymology, 173, 841-841 (1989)
Parker, L.
Methods in Enzymology, 190, 480-480 (1990)
Alan Moreira de Araujo et al.
Cells, 7(8) (2018-08-01)
Hepatocytes may rupture after a drug overdose, and their intracellular contents act as damage-associated molecular patterns (DAMPs) that lead to additional leukocyte infiltration, amplifying the original injury. Necrosis-derived DNA can be recognized as a DAMP, activating liver non-parenchymal cells (NPCs).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service