Skip to Content
Merck
All Photos(1)

Documents

COLLA-RO

Roche

Collagenase A

from Clostridium histolyticum

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352204

biological source

Clostridium histolyticum

Quality Level

sterility

non-sterile

form

lyophilized

specific activity

>0.15 U/mg lyophilizate (Collagenase activity)

packaging

pkg of 100 mg (10103578001)
pkg of 2.5 g (11088793001)
pkg of 500 mg (10103586001)

manufacturer/tradename

Roche

concentration

1 mg/mL (0.1%, w/v)

color

brown

optimum pH

6.0-8.0

solubility

water: soluble

suitability

suitable for PCR

NCBI accession no.

UniProt accession no.

application(s)

cell analysis
life science and biopharma
sample preparation

foreign activity

Clostripain 9.489 U/mg
Proteases 6.067 U/mg (Azocoll )
Trypsin 0.144 U/mg (with BAEE)

storage temp.

2-8°C

Gene Information

Clostridium histolyticum ... COLA_1(1668506084)

General description

Collagenase is a protease which cleaves the triple-helical protein called collagen. Clostridium histolyticum produces six types of collagenases namely, α, β, γ, δ, ε and λ. These collagenases are active against connective tissue and show more reactivity towards gelatin than collagen. Collagenase obtained from Clostridium histolyticum has a very strong activity, as it digests collagen from both ends, at temperatures as low as 4-10 °C.

Specificity

Collagenase degrades native collagen. Clostripain, trypsin-like enzymes, and neutral proteases also degrade other proteins.
Enzyme activity:
Collagenase activity: >0.15U/mg (according to Wünsch) (+25°C, 4-phenyl-azobenzyl-oxycarbonyl-Pro-Leu-Gly-Pro-D-Arg as the substrate)
Contaminating enzyme activities: trypsin, clostripain, and neutral proteolytic activity
Collagenase A has a balanced ratio of enzyme activities.

Application

Collagenase from C. histolyticum is widely used for the:
  • Disaggregation of many types of tissues (e.g., lung, heart, muscle, bone, adipose tissue, liver, kidney, cartilage, mammary gland, placentae, blood vessels, brain, tumors, umbilical cord)
  • preparation of single-cell suspensions for the establishment of primary cell culture systems. Collagenase A is recommended when yield, viability, and functionality are important.
  • preparation of cells from many types of tissue, such as hepatocytes, adipocytes, pancreatic islets, epithelial cells, muscle cells, endothelial cells, etc. However, the suitability of each lot of the enzyme for disruption of a particular tissue should be determined empirically.

Unit Definition

Contents
Lyophilizate, nonsterile
Unit Definition: Collagenase from Roche is assayed in Wünsch units (1 μmol of product formed per minute at +25 °C with Wünsch substrate).
Frequently, collagenase activities are given in Mandl units (1 μmol leucine liberated from collagen in 5 hours at +37 °C).
Unfortunately, there is no consistent conversion factor between the two units of activity, since the Mandl unit depends, in part, on the concentration of contaminating proteases in the collagenase preparation, an indefinable variable. A purer collagenase preparation would actually give a lower specific activity in Mandl units than a crude preparation. Clostridium preparations typically give conversion factors of approximately 1:1800 (e.g., a particular lot of Clostridium collagenase contained approximately 0.15 Wünsch U/mg and 250 Mandl U/mg).

Preparation Note

Activator: Ca2+
Inhibitors:
Collagenase inhibitors: EDTA, EGTA, Cys, His, DTT, 2-mercapto-ethanol
Collagenase is not inhibited by serum.
Clostripain inhibitors: TLCK
Trypsin inhibitors: aprotinin, trypsin inhibitor (egg white, soybean), serum
Working concentration: 0.5 to 2.5 mg/ml
Storage conditions (working solution): -15 to -25 °C
Roche recommends reconstituting only the amount of lyophilizate needed for immediate use. The reconstituted solution can be stored at -15 to -25 °C for up to one week. Avoid repeated freezing and thawing since activity decreases after reconstitution.

Reconstitution

Reconstitution in any balanced salt solution (e.g., HBSS)

Storage and Stability

Keep container tightly closed in a dry and well-ventilated place

Other Notes

For life science research only. Not for use in diagnostic procedures.
Shipping conditions may vary

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rajesh K Mistry et al.
The Journal of biological chemistry, 291(4), 1774-1788 (2015-12-02)
The gasotransmitter, hydrogen sulfide (H2S) is recognized as an important mediator of endothelial cell homeostasis and function that impacts upon vascular tone and blood pressure. Cystathionine-γ-lyase (CSE) is the predominant endothelial generator of H2S, and recent evidence suggests that its
Matthew Kennedy et al.
BMJ open respiratory research, 5(1), e000321-e000321 (2018-10-03)
The degree to which bacteria in the human respiratory tract are aerosolised by individuals is not established. Building on our experience sampling bacteria exhaled by individuals with pulmonary tuberculosis using face masks, we hypothesised that patients with conditions frequently treated
Henning Thomasen et al.
Investigative ophthalmology & visual science, 56(8), 4460-4469 (2015-07-22)
Until now, no epithelial cell line from conjunctival squamous cell carcinoma (SCC), to our knowledge, has existed; therefore, the establishment of a model cell line would be a useful tool for further studies. In particular, the phenotypic and molecular characterization
Exhaled Pneumocystis jirovecii output and detection of asymptomatic exhalation by facemask sampling in HIV-uninfected, immunocompromised patients
Abdulwhhab MT, et al.
Journal of Hospital Infection, 132, 20-27 (2023)
Jonathan A Cohen et al.
Cell, 178(4), 919-932 (2019-07-30)
Cutaneous TRPV1+ neurons directly sense noxious stimuli, inflammatory cytokines, and pathogen-associated molecules and are required for innate immunity against some skin pathogens. Important unanswered questions are whether TRPV1+ neuron activation in isolation is sufficient to initiate innate immune responses and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service