Skip to Content
Merck
All Photos(1)

Key Documents

C160

Sigma-Aldrich

CGP-42112A

≥95%, synthetic, solid

Synonym(s):

Nα-Nicotinoyl-Tyr-(Nα-Cbz-Arg)-Lys-His-Pro-Ile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C52H69N13O11
CAS Number:
Molecular Weight:
1052.18
Beilstein:
8184948
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic

Quality Level

Assay

≥95%

form

solid

color

white

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@@H]1CCCN1C(=O)[C@H](Cc2c[nH]cn2)NC(=O)[C@H](CCCCNC(=O)[C@H](CCCNC(N)=N)NC(=O)OCc3ccccc3)NC(=O)[C@H](Cc4ccc(O)cc4)NC(=O)c5cccnc5)C(O)=O

InChI

1S/C52H69N13O11/c1-3-32(2)43(50(73)74)64-48(71)42-17-11-25-65(42)49(72)41(27-36-29-56-31-59-36)62-46(69)39(60-47(70)40(26-33-18-20-37(66)21-19-33)61-44(67)35-14-9-22-55-28-35)15-7-8-23-57-45(68)38(16-10-24-58-51(53)54)63-52(75)76-30-34-12-5-4-6-13-34/h4-6,9,12-14,18-22,28-29,31-32,38-43,66H,3,7-8,10-11,15-17,23-27,30H2,1-2H3,(H,56,59)(H,57,68)(H,60,70)(H,61,67)(H,62,69)(H,63,75)(H,64,71)(H,73,74)(H4,53,54,58)/t32-,38-,39-,40-,41-,42-,43-/m0/s1

InChI key

UXGNARZDONUMMK-LRMQDCNJSA-N

Gene Information

human ... AGTR2(186)
rat ... AGTR2(11609)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Tyr-Z-Arg-Lys-His-Pro-Ile

Application

CGP-42112A has been used:
  • in the isolation of RNA from fibroblast-like synoviocytes from rheumatoid arthritis patients (RA-FLS)
  • to study its effects on the ERK2 phosphorylation in the cardiomyoblast cell line
  • as a stimulator of angiotensin II type 2 (AT2) receptor to study its effects on human regional hemodynamic responses mediated by AT2 receptors

Biochem/physiol Actions

CGP-42112A is a potent angiotensin II type 2 receptor (AT2R) agonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anu Vaajanen et al.
Investigative ophthalmology & visual science, 49(6), 2557-2562 (2008-01-29)
In the present study the effects of exogenous angiotensin II and its breakdown metabolite angiotensin (1-7) on the intraocular pressure (IOP) and on aqueous humor dynamics in normotensive rabbit eye were evaluated. Male New Zealand White rabbits with normal IOP
C Sumners et al.
Proceedings of the National Academy of Sciences of the United States of America, 88(17), 7567-7571 (1991-09-01)
Both neurons and astrocytes contain specific receptors for angiotensin II (AII). We used selective ligands for the AT1 and AT2 types of AII receptors to investigate the expression of functional receptor subtypes in astrocyte cultures and neuron cultures from 1-day-old
Nancy J Hong et al.
Hypertension (Dallas, Tex. : 1979), 60(3), 765-769 (2012-07-11)
NO reduces NaCl absorption by thick ascending limbs (TALs) by inhibiting the Na/K/2Cl cotransporter (NKCC2). We have shown that NO-induced inhibition of Na transport is reduced in Dahl salt-sensitive rat (SS) TALs. Angiotensin II increases NO production in TALs via
Liping Zhu et al.
American journal of physiology. Heart and circulatory physiology, 302(12), H2553-H2559 (2012-04-24)
ANG II type 2 receptors (AT(2)R) elicit cardioprotective effects in part by stimulating the release of kinins; however, the mechanism(s) responsible have not been fully explored. We demonstrated previously that overexpression of AT(2)R increased expression of prolylcarboxypeptidase (PRCP; a plasma
Claudia A McCarthy et al.
Hypertension (Dallas, Tex. : 1979), 60(6), 1531-1537 (2012-10-24)
We have demonstrated previously that pretreatment with an angiotensin II type 2 receptor (AT(2)R) agonist is neuroprotective against a subsequent stroke independent of any changes in blood pressure. Therefore, in the current study, we have examined the potential neuroprotective effect

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service