Skip to Content
Merck
All Photos(1)

Key Documents

CASP3C

Sigma-Aldrich

Caspase 3 Assay Kit, Colorimetric

Synonym(s):

Colorimetric Caspase-3 Activity Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84
Pricing and availability is not currently available.

usage

sufficient for 1,000 multiwell tests (in 96-well multiwell plates)
sufficient for 100 standard tests (1 ml)

Quality Level

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... CASP3(836)

Related Categories

General description

Caspase 3 Colorimetric Assay Kit provides the reagents needed for a quick and efficient detection of caspase 3 activity in cell lysates and in purified preparations of caspase 3.

The Caspase 3 Colorimetric Assay Kit is based on the hydrolysis of acetyl-Asp-Glu-Val-Asp p-nitroanilide (Ac-DEVD-pNA) by caspase 3, resulting in the release of the p-nitroaniline (pNA) moiety. p-Nitroaniline is detected at 405 nm (εmM=10.5). The concentration of the pNA released from the substrate is calculated from either the absorbance values at 405 nm or from a calibration curve prepared with pNA standards (pNA standard included with the kit).

Application

The assay can be performed (a) in 1 mL volume and measured using a spectrophotometer, or (b) in 100 µL volume in a 96-well plate using an ELISA reader.

Unit Definition

One unit of caspase 3 will cleave 1.0 μmol of Ac-DEVD-pNA per min at pH 7.4 at 25 °C.

Other Notes

Use 17 megohm water only

Kit Components Only

Product No.
Description

  • Water (17-18 megaOhm) 125 mL

Kit Components Also Available Separately

Product No.
Description
SDS

  • A0835Ac-DEVD-CHO inhibitor .5 mgSDS

  • A2559Ac-DEVD-pNA substrate 15 mgSDS

  • p-Nitroaniline standard 1 mg

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point(F)

415.4 °F - closed cup

Flash Point(C)

213 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alaa Jameel A Albarakati et al.
Molecular biology reports, 47(4), 2591-2603 (2020-03-08)
Lead (Pb) is one of the most common heavy metal pollutants affecting living organisms. It induces nephrotoxicity with significant alterations in renal structure and function. Luteolin (LUT) a flavonoid present in various plant products is well known for exhibiting numerous
Gadah Albasher et al.
Oxidative medicine and cellular longevity, 2020, 2963020-2963020 (2020-03-28)
Organophosphorus insecticides including chlorpyrifos (CPF) are mainly used for agriculture, household, and military purposes; their application is associated with various adverse reactions in animals and humans. This study was conducted to evaluate the potential neuroprotective effect of red beetroot methanolic
S C Masters et al.
The Journal of biological chemistry, 276(48), 45193-45200 (2001-09-29)
The 14-3-3 proteins are a family of highly conserved eukaryotic regulatory molecules that play important roles in many biological processes including cell cycle control and regulation of cell death. They are able to carry out these effects through binding and
Wafa A Al-Megrin et al.
Frontiers in physiology, 11, 64-64 (2020-03-03)
The kidney is among the metabolic organs most susceptible to injury, particularly following exposure to xenobiotics and heavy metals. We aimed to explore the potential protective impacts of coenzyme Q10 (CoQ10) on lead acetate (PbAc)-induced nephrotoxicity in rats. Four experimental
Afrah F Alkhuriji et al.
Animals : an open access journal from MDPI, 10(4) (2020-04-03)
Listeria monocytogenes is a psychrophilic bacterium, which causes widespread zoonosis in the natural environment, and mainly affects goat, sheep, and cattle herds. Recently, we predicted that it can be transmitted through food. It causes listeriosis, a severe infectious disease, which

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service