Skip to Content
Merck
All Photos(1)

Documents

276685

Sigma-Aldrich

Methacrylic anhydride

contains 2,000 ppm topanol A as inhibitor, ≥94%

Synonym(s):

Methacrylic acid anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[H2C=C(CH3)CO]2O
CAS Number:
Molecular Weight:
154.16
Beilstein:
1761982
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

≥94%

form

liquid

contains

2,000 ppm topanol A as inhibitor

refractive index

n20/D 1.453 (lit.)

bp

87 °C/13 mmHg (lit.)

density

1.035 g/mL at 25 °C (lit.)

SMILES string

CC(=C)C(=O)OC(=O)C(C)=C

InChI

1S/C8H10O3/c1-5(2)7(9)11-8(10)6(3)4/h1,3H2,2,4H3

InChI key

DCUFMVPCXCSVNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methacrylic anhydride is an organic compound derived from methacrylic acid, which belongs to the class of acrylic acids. It is primarily employed as a monomer in the production of polymers, particularly poly(methacrylic acid) and its copolymers. These polymers find applications in a wide range of industries, including bio-renewable resins, pH-sensitive hydrogels, thermosets, coatings, adhesives, and plastics, due to their excellent chemical and thermal stability. In the polymerization of methacrylic anhydride, the most common inhibitor used is Topanol A. It is typically added in small quantities (2,000 ppm) to hinder undesirable side reactions during the polymerization, ensuring controlled and high-quality polymer formation.

Application

Methacrylic anhydride can be used as a starting material to synthesize:
  • Methacrylated chondroitin sulfate pH-sensitive hydrogels by copolymerization reaction. These hydrogels can be used in drug delivery systems.
  • High-performance lignin-based thermosets.
  • Gel polymer electrolyte, which is then integrated with the cathode to form high-performance lithium-ion batteries.
Methacrylic anhydride can be also used as a precursor to synthesize bio-renewable resins for composite applications. For example, it can be used to prepare soybean oil and a eugenol-based resin system. This resin possesses high thermal stability, and good mechanical properties, which makes it a suitable matrix for the pultrusion process.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Notes-Acrylic and Methacrylic Anhydrides.
Brotherton T, et al.
The Journal of Organic Chemistry, 26(4), 1283-1284 (1961)
Cyclopolymerization. IV. Structure of polymethacrylic anhydride and kinetics of polymerization of methacrylic anhydride.
Smets G, et al.
Journal of Polymer Science: Part A, General Papers, 2(11), 4825-4834 (1964)
A I Van Den Bulcke et al.
Biomacromolecules, 1(1), 31-38 (2001-11-17)
Dynamic shear oscillation measurements at small strain were used to characterize the viscoelastic properties and related differences in the molecular structure of hydrogels based on gelatin methacrylamide. Gelatin was derivatized with methacrylamide side groups and was subsequently cross-linked by radical
G Bahcecioglu et al.
Biomaterials, 218, 119361-119361 (2019-07-25)
A PCL/hydrogel construct that would mimic the structural organization, biochemistry and anatomy of meniscus was engineered. The compressive (380 ± 40 kPa) and tensile modulus (18.2 ± 0.9 MPa) of the PCL scaffolds were increased significantly when constructs were printed with a shifted design and circumferential
Avathamsa Athirasala et al.
Scientific reports, 7(1), 3323-3323 (2017-06-14)
The requirement for immediate vascularization of engineered dental pulp poses a major hurdle towards successful implementation of pulp regeneration as an effective therapeutic strategy for root canal therapy, especially in adult teeth. Here, we demonstrate a novel strategy to engineer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service