Skip to Content
Merck
All Photos(1)

Key Documents

C0494

Sigma-Aldrich

Cambinol

≥97% (HPLC), white, powder

Synonym(s):

5-(2-Hydroxynaphthalen-1-ylmethyl)-6-phenyl-2-thioxo-2,3-dihydro-1H-pyrimidin-4-one, 5-[(2-hydroxy-1-naphthyl)methyl]-2-mercapto-6-phenyl-4(3H)-Pyrimidinone, NSC-1125476, Tetrahydro-5-[(2-hydroxy-1-naphthalenyl)methyl]-6-phenyl-2-thioxo-4(1H)-Pyrimidinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H16N2O2S
CAS Number:
Molecular Weight:
360.43
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white

solubility

DMSO: ≥10 mg/mL

storage temp.

room temp

SMILES string

Oc1ccc2ccccc2c1CC3=C(NC(=S)NC3=O)c4ccccc4

InChI

1S/C21H16N2O2S/c24-18-11-10-13-6-4-5-9-15(13)16(18)12-17-19(14-7-2-1-3-8-14)22-21(26)23-20(17)25/h1-11,24H,12H2,(H2,22,23,25,26)

InChI key

RVNSQVIUFZVNAU-UHFFFAOYSA-N

Application

Cambinol has been used:
  • as an inhibitor of sirutins (SIRT1 and 2) in hepatocarcinoma cell lines to test its effect on cellular viability and colony formation
  • as a SIRT1 and SIRT2 inhibitor to test its effect on frizzled 7 (FZD7) expression in the breast cancer cells MDA-MB-231 and T-47D cells
  • as a sirutin inhibitor in L929 cells to test its protective effect against tumor necrosis factor (TNF)-induced cell death

Biochem/physiol Actions

Cambinol is a Sirtuin (Human Silent Information Regulator) Type 1/2 Inhibitor.
Cambinol is a Sirtuin (Human Silent Information Regulator) Type 1/2 Inhibitor. Sirtuins are structurally and mechanistically unrelated to HDACs Class 1 & 2 deacetylases but share many protein targets. HDAC is a large complex enzyme family involved in epigenetic-control of gene expression. Sirt2 has an IC50 equal to 59 μM.
Cambinol, a β-naphthol compound, elicits antitumor activity and impairs the mitogen-activated protein kinase (MAPK) signaling. It has ability to inhibit the secretion of inflammatory cytokines including the tumor necrosis factor and interleukins. Cambinol can be a potential therapeutic agent for treating inflammatory diseases. It also aids protection in events of endotoxic and septic shock.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frizzled 7 expression is positively regulated by SIRT1 and beta-catenin in breast cancer cells
Simmons Jr GE, et al.
PLoS ONE, 9(6), e98861-e98861 (2014)
The sirtuin inhibitor cambinol impairs MAPK signaling, inhibits inflammatory and innate immune responses and protects from septic shock
Lugrin J, et al.
Biochimica et Biophysica Acta - Molecular Cell Research, 1833(6), 1498-1510 (2013)
Inhibition of sirtuins 1 and 2 impairs cell survival and migration and modulates the expression of P-glycoprotein and MRP3 in hepatocellular carcinoma cell lines
Ceballos MP, et al.
Toxicology Letters, 289(6), 63-74 (2018)
Marie Gamal et al.
Current issues in molecular biology, 45(6), 5005-5017 (2023-06-27)
Liver cancer is a prevalent form of cancer worldwide. While research has shown that increasing sphingomyelin (SM) hydrolysis by activating the cell surface membrane-associated neutral sphingomyelinase 2 (nSMase2) can control cell proliferation and apoptosis, the role of total glutathione depletion
Intracellular nicotinamide adenine dinucleotide promotes TNF-induced necroptosis in a sirtuin-dependent manner
Preyat N, et al.
Cell Death and Differentiation, 23(1), 29-40 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service