Skip to Content
Merck
All Photos(1)

Documents

A8231

Sigma-Aldrich

AGK2

≥97% (HPLC), powder

Synonym(s):

2-Cyano-3-[5-(2,5-dichlorophenyl)-2-furanyl]-N-5-quinolinyl-2-propenamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H13Cl2N3O2
CAS Number:
Molecular Weight:
434.27
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97% (HPLC)

form

powder

solubility

DMSO: 2 mg/mL, clear (warmed)
H2O: insoluble

storage temp.

room temp

SMILES string

Clc1ccc(Cl)c(c1)-c2ccc(\C=C(/C#N)C(=O)Nc3cccc4ncccc34)o2

InChI

1S/C23H13Cl2N3O2/c24-15-6-8-19(25)18(12-15)22-9-7-16(30-22)11-14(13-26)23(29)28-21-5-1-4-20-17(21)3-2-10-27-20/h1-12H,(H,28,29)/b14-11+

InChI key

SVENPFFEMUOOGK-SDNWHVSQSA-N

Application

AGK2 is a SIRT2 inhibitor. AGK2 has been used in a study to determine that SIRT2 inhibition induces cell death and decreases the intracellular ATP level. AGK2 also rescues dopamine neurons from α-synuclein toxicity in Parkinson′s disease models.

Biochem/physiol Actions

AGK2 is a SIRT2 inhibitor. AGK2 rescues dopamine neurons from α-synuclein toxicity in Parkinson′s disease models. IC50 for SIRT2 = 3.5 uM. AGK2 is >15-fold more selective for SIRT2 than SIRT1 and SIRT3. AGK2 may be the most selective SIRT2 inhibitor available.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hai-Bo Yu et al.
International journal of medical sciences, 15(12), 1356-1364 (2018-10-03)
Sirtuin 2 (SIRT2) is a nicotinamide adenine dinucleotide (NAD +)-dependent class III histone deacetylase. We have reported that HBx (hepatitis B virus X protein)-elevated SIRT2 promotes HBV replication and hepatocarcinogenesis. However, the potential anti-HBV effect of AGK2, a selective inhibitor
David T She et al.
Molecular neurobiology, 55(12), 9188-9203 (2018-04-15)
Sirtuin 2 (SIRT2) is a family member of nicotinamide adenine dinucleotide (NAD+)-dependent deacetylases which appears to have detrimental roles in an array of neurological disorders such as Parkinson's disease (PD) and Huntington's disease (HD). In light of the recently emerging
Lisa M Giammona et al.
Experimental hematology, 37(11), 1340-1352 (2009-09-01)
Megakaryocytic cells (Mks) undergo endomitosis and become polyploid. Mk ploidy correlates with platelet production. We previously showed that nicotinamide (NIC) greatly increases Mk ploidy in cultures of human mobilized peripheral blood CD34(+) cells. This study aims to examine the generality
G L Garrote et al.
The Journal of dairy research, 68(4), 639-652 (2002-04-04)
Chemical and microbiological composition of four Argentinean kefir grains from different sources as well as characteristics of the corresponding fermented milk were studied. Kefir grains CIDCA AGK1, AGK2 and AGK4 did not show significant differences in their chemical and microbiological
Sapna Singh et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 36(8), 6159-6171 (2015-03-22)
Sirtuins belong to the family of class III histone deacetylases; its role in neoplasia is controversial as both tumor-suppressive and promoting functions have been reported. There are very few reports available, where expressions of sirtuin isoforms are comprehensively analyzed during

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service