Skip to Content
Merck

EPI006

Histone Deacetylase 8 (HDAC8) Activity Assay Kit

100 assays in 96 well plates

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

EC Number:
NACRES:
NA.41
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


usage

100 assays in 96 well plates

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... HDAC8(55869)
mouse ... HDAC8(70315)

General description

Histone deacetylases (HDACs) are a large family of enzymes that remove acetyl groups from histone proteins. Site specific histone acetylation and deacetylation have been shown to activate or repress eukaryotic gene transcription, respectively, and as a consequence, it plays a crucial role in mammalian development and disease. HDACs are involved in important biological activities, such as cell differentiation, proliferation, apoptosis, and senescence.

With Sigma′s HDAC8 Activity Assay Kit, HDAC8 present in a test sample will act with the supplied Developer, to deacetylate and then cleave the HDAC8 Substrate (R-H-K(Ac)-K(Ac)-AFC). This activity will release the quenched fluorescent group, AFC, which can be detected at Em/Ex = 380/500 nm. Trichostatin A is an HDAC inhibitor included in the kit to verify HDAC8 activity. The kit provides a rapid, simple, sensitive, and reliable test. It is suitable for either individual tests or high throughput assays, from nuclear extracts, purified, or immunoprecipitated HDAC8, and from native, recombinant, or genetically modified HDAC8.
HDAC8 (histone deacetylase 8) is a class I enzyme, that has 377 residues and it lies between class I and class II HDACs. This gene is located on X chromosome. It consists of a single α/β domain, in which eight parallel stranded β sheets are sandwiched between 13 α helices.

Biochem/physiol Actions

HDAC8 (histone deacetylase 8), with the help of PKA (cyclic AMP-dependent protein kinase A) plays a major role in confirming the acetylation state of histones. This protein may also plays an important role in acute myeloid leukemia (AML). Suppression of HDAC8 by RNA interference stops the growth of lung, colon, and cervical cancer cell lines.

Features and Benefits

  • Simple, sensitive, and reliable assay
  • Utilizes fluorometric methods
  • Sample type: cell and tissue lysates, plasma and serum, other biological fluids
  • Species reactivity: mammalian
  • Suitable for individual tests or high throughput assays and kinetic studies
  • Suitable for high throughput measurement of HDAC8 activity in purified, immunoprecipitated, and recombinant or genetically modified HDAC8 samples
  • Convenient 96-well microplate format


Storage Class

10 - Combustible liquids

WGK

WGK 3

Flash Point (°F)

188.6 °F - closed cup

Flash Point (°C)

87 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library