Skip to Content
Merck

Skip To

D3900

Desipramine hydrochloride

≥98% (TLC), powder, imipramine metabolite

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C18H22N2 · HCl
CAS Number:
Molecular Weight:
302.84
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352116
EC Number:
200-373-1
MDL number:
Assay:
≥98% (TLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Desipramine hydrochloride, ≥98% (TLC), powder

Quality Segment

assay

≥98% (TLC)

form

powder

color

white to off-white

solubility

H2O: 50 mg/mL

originator

Sanofi Aventis

storage temp.

2-8°C

SMILES string

Cl[H].CNCCCN1c2ccccc2CCc3ccccc13

InChI

1S/C18H22N2.ClH/c1-19-13-6-14-20-17-9-4-2-7-15(17)11-12-16-8-3-5-10-18(16)20;/h2-5,7-10,19H,6,11-14H2,1H3;1H

InChI key

XAEWZDYWZHIUCT-UHFFFAOYSA-N

Gene Information

human ... SLC6A2(6530)

General description

Desipramine, a metabolite of imipramine and iofepramine is an active tricyclic antidepressant.

Application

Desipramine hydrochloride has been used:
  • to prevent norepinephrine neuron degeneration and to maximize uptake into DA neurons
  • to protect unselective damage to the noradrenergic system[1]
  • as a norepinephrine re-uptake inhibitor to treat rats for lesioning

Biochem/physiol Actions

Desipramine is used to treat depression. It is also used to manage enuresis in children. Desipramine helps to eliminate the pain triggered by diabetic neuropathy.
Tricyclic antidepressant that is a more potent inhibitor of the norepinephrine transporter (Ki = 4 nM) than the serotonin transporter (Ki = 61 nM).

Features and Benefits

This compound is featured on the Biogenic Amine Transporters page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Sanofi Aventis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Disclaimer

Hygroscopic

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1172006D0500000BP931
form

powder

form

-

form

-

form

solid

assay

≥98% (TLC)

assay

-

assay

-

assay

-

Quality Level

100

Quality Level

-

Quality Level

-

Quality Level

-

storage temp.

2-8°C

storage temp.

-

storage temp.

-

storage temp.

2-8°C

solubility

H2O: 50 mg/mL

solubility

-

solubility

-

solubility

-

originator

Sanofi Aventis

originator

-

originator

-

originator

-


Still not finding the right product?

Explore all of our products under Desipramine hydrochloride


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
D3900-1G04061835253166
D3900-25G04061835253173
D3900-5G04061833563694

Questions

Reviews

No rating value

Active Filters