Skip to Content
Merck
All Photos(6)

Documents

A9528

Sigma-Aldrich

Amphotericin B solubilized

powder, suitable for cell culture, BioXtra

Synonym(s):

Amphotericin B from Streptomyces sp., Fungizone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H73NO17
CAS Number:
Molecular Weight:
924.08
Beilstein:
78342
MDL number:
UNSPSC Code:
12352207
eCl@ss:
34010400
PubChem Substance ID:
NACRES:
NA.76

product name

Amphotericin B solubilized, powder, γ-irradiated, BioXtra, suitable for cell culture

Quality Level

sterility

γ-irradiated

product line

BioXtra

form

powder

technique(s)

cell culture | mammalian: suitable

antibiotic activity spectrum

fungi
yeast

Mode of action

cell membrane | interferes

storage temp.

2-8°C

SMILES string

C[C@H]1O[C@@H](O[C@@H]2C[C@@H]3O[C@](O)(C[C@@H](O)C[C@@H](O)[C@H](O)CC[C@@H](O)C[C@@H](O)CC(=O)O[C@@H](C)[C@H](C)[C@H](O)[C@@H](C)\C=C\C=C\C=C\C=C\C=C\C=C\C=C\2)C[C@H](O)[C@H]3C(O)=O)[C@@H](O)[C@@H](N)[C@@H]1O

InChI

1S/C47H73NO17/c1-27-17-15-13-11-9-7-5-6-8-10-12-14-16-18-34(64-46-44(58)41(48)43(57)30(4)63-46)24-38-40(45(59)60)37(54)26-47(61,65-38)25-33(51)22-36(53)35(52)20-19-31(49)21-32(50)23-39(55)62-29(3)28(2)42(27)56/h5-18,27-38,40-44,46,49-54,56-58,61H,19-26,48H2,1-4H3,(H,59,60)/b6-5+,9-7+,10-8+,13-11+,14-12+,17-15+,18-16+/t27-,28-,29-,30+,31+,32+,33-,34-,35+,36+,37-,38-,40+,41-,42+,43+,44-,46-,47+/m0/s1

InChI key

APKFDSVGJQXUKY-INPOYWNPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: polyene

Application

Amphotericin B is a polyene antifungal antibiotic from Streptomyce sp. It has a high affinity for sterols, especially ergosterols, of both fungal and bacterial cell membranes. Normal usage in maintenance of antifungal cell cultures is 2.5 mg/L with penicillin and streptomycin used in the medium.

Biochem/physiol Actions

Amphotericin B binds to sterols, forming pores in the membrane, and causing small molecules to leak out. The product is most effective against fungi and yeast.

Packaging

Solubilized Amphotericin B is packaged in amber glass bottle, with the contents inside an inserted fused cone.

Components

This product is a colloidal suspension of Amphotericin B, using deoxycholate as the solubilizing agent. The formulation is ~45% Amphotericin B and 35% sodium deoxycholate in sodium phosphate and sodium chloride.

Caution

Amphotericin B remains active for 3 days in culture at 37°C. At -20°C, and with protection from air and light, the product remains active for 5 years.

Preparation Note

Amphotericin B is soluble at 19.6-20.4 mg/ml in water, at natural pH without heating, giving a yellowish clear to slightly hazy solution.
Aqueous solutions cannot be sterile filtered due to poor solubility. If reconstituted in sterile water, there is no need to filter sterilize.

It is also soluble in DMSO (30-40 mg/mL) and in dimethylformamide (2-4 mg/mL).

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Product is sensitive to light and moisture. Keep in a dry place.
The actual solid weight of the packaged product is calculated according to the UV purity.

Kit Components Only

Product No.
Description

  • Amphotericin B ~ 45 %

  • Sodium deoxycholate ~ 35 %

  • Sodium phosphate balance

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Derayvia Grimes et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(39), 24403-24414 (2020-09-16)
Calcium signals are initiated in immune cells by the process of store-operated calcium entry (SOCE), where receptor activation triggers transient calcium release from the endoplasmic reticulum, followed by opening of plasma-membrane calcium-release activated calcium (CRAC) channels. ORAI1, ORAI2, and ORAI3
Devin Tran et al.
Cornea, 38(7), 901-904 (2019-04-19)
Fungal infections in lamellar keratoplasty are a growing concern. Optisol-GS does not contain an antifungal agent and supplementation with 0.255 μg/mL Amphotericin B (AmpB) has been considered. This study tested the ability of 0.255 μg/mL AmpB in Optisol-GS to eliminate
Etienne Loire et al.
Heliyon, 6(10), e05146-e05146 (2020-10-22)
Contagious caprine pleuropneumonia is an infectious and contagious disease affecting goats and wildlife ruminants, mostly in Africa and Asia. It is caused by a mycoplasma, Mycoplasma capricolum susbp. capripneumoniae, which is very fastidious. This may be the reason why there
Ziying Yan et al.
Current protocols in microbiology, 58(1), e107-e107 (2020-07-09)
Human bocavirus 1 (HBoV1) is a small DNA virus that belongs to the Bocaparvovirus genus of the Parvoviridae family. HBoV1 is a common respiratory pathogen that causes mild to life-threatening acute respiratory tract infections in children and immunocompromised individuals, infecting
Malav S Trivedi et al.
The Journal of nutritional biochemistry, 25(10), 1011-1018 (2014-07-16)
Dietary interventions like gluten-free and casein-free diets have been reported to improve intestinal, autoimmune and neurological symptoms in patients with a variety of conditions; however, the underlying mechanism of benefit for such diets remains unclear. Epigenetic programming, including CpG methylation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service