Skip to Content
Merck
All Photos(1)

Key Documents

N9150

Sigma-Aldrich

Nystatin

ready made solution, suitable for cell culture

Synonym(s):

Nystatin, Fungicidin, Mycostatin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352207
NACRES:
NA.76
Pricing and availability is not currently available.

Product Name

Nystatin Ready made solution, suitable for cell culture

biological source

Streptomyces noursei

form

solution

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@H]([C@H](C)[C@H](C)O1)[C@@H](C)/C=C/C=C/CC/C=C/C=C/C=C/C=C/[C@H](O[C@@H]2O[C@H](C)[C@@H](O)[C@H](N)[C@@H]2O)C[C@@]3([H])[C@H](C(O)=O)[C@@H](O)C[C@](O)(O3)C[C@@H](O)[C@H](O)CC[C@@H](O)C[C@@H](O)C[C@@H](O)CC1=O

InChI

1S/C47H75NO17/c1-27-17-15-13-11-9-7-5-6-8-10-12-14-16-18-34(64-46-44(58)41(48)43(57)30(4)63-46)24-38-40(45(59)60)37(54)26-47(61,65-38)25-36(53)35(52)20-19-31(49)21-32(50)22-33(51)23-39(55)62-29(3)28(2)42(27)56/h5-6,8,10-18,27-38,40-44,46,49-54,56-58,61H,7,9,19-26,48H2,1-4H3,(H,59,60)/b6-5+,10-8+,13-11+,14-12+,17-15+,18-16+/t27-,28+,29-,30+,31+,32+,33+,34-,35+,36+,37-,38-,40+,41-,42+,43+,44-,46-,47+/m0/s1

InChI key

VQOXZBDYSJBXMA-QEKUPDCNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nystatin is a fungistatic and fungicidal polyene antibiotic. It binds to ergosterol which increases cell membrane permeability in sensitive fungi.[1] Nystatin is mainly clinically active against Candida species.[2] However, it is also reported to be effective against Aspergillus, Coccidioides immitis, Cryptococcus neoformans, Histoplasma capsulatum, Blastomyces dermatidis, and other yeasts and fungi.[3] Nystatin has also been used to enrich mutants by negative selection in yeasts.[4] The minimum inhibitory concentration (MIC) of Nystatin to Saccharomyces cerevisiae was reported to be 3μg/ml [5] while for clinical isolates of Candida albicans the MIC was reported to be 4-16μg/ml.[6]

Biochem/physiol Actions

Mode of Action: Nystatin acts by binding to ergosterols in fungal cell membranes, increasing permeability of the membrane, and creating nystatin/ergosterol based ion channels. It also functions as a lipid raft-inhibiting reagent and as membrane associated cholesterol.

Antimicrobial spectrum: Nystatin acts against fungi, yeasts and molds.

Features and Benefits

  • Nystatin ready made solution is a fully solubilized formulation of Nystatin with potency of 8000-13000 units/ml according to USP standard.
  • Nystatin ready made solution is 0.2μm filtered, endotoxin.
  • The suggested working concentration of Nystatin ready made solution is 30 units/ml.
  • The ready made solution is light sensitive. It is recommended to store the solution in the dark.
  • It is recommended to avoid freeze thaw cycles of this product. Therefore, it is recommended to aliquot and store at -20°C.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martindale The Extra Pharmacopoeia
Reynolds, J. E. F.
Martindale: The Extra Pharmacopoeia, 432-433 null
Polyene toxicity in renal medulla: injury mediated by transport activity
Brezis, M., et al.
Science, 224, 66-68 (1984)
Antifungal drug susceptibility of Candida albicans
Bii CC, Ouko TT, Amukoye E, Githinji LW
East African Medical Journal, 79(3), 143-145 (2002)
Clinical Practice Guideline for the Management of Candidiasis: 2016 Update by the Infectious Diseases Society of America. Clinical infectious diseases : an official publication of the Infectious Diseases Society of America
Pappas PG, Kauffman CA, Andes DR, et al.
Infectious Diseases, 62, el-50 (2016)
Polyene macrolide derivatives. III
Bonner, D. P., Mechlinski, W., and Schaffner, C. P.
The Journal of Antibiotics, 25 (4), 261-262 (1972)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service