Skip to Content
Merck
All Photos(6)

Key Documents

A4888

Sigma-Aldrich

Amphotericin B from Streptomyces sp.

~80% (HPLC), powder

Synonym(s):

LNS-AmB, NSC 527017, Fungizone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H73NO17
CAS Number:
Molecular Weight:
924.08
Beilstein:
78342
EC Number:
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.85
Pricing and availability is not currently available.

biological source

Streptomyces sp.

Quality Level

Assay

~80% (HPLC)

form

powder

color

yellow to orange

solubility

DMF: 2-4 mg/mL
DMSO: 30-40 mg/mL
H2O: insoluble
water: insoluble

antibiotic activity spectrum

fungi
yeast

Mode of action

cell membrane | interferes

storage temp.

2-8°C

SMILES string

C[C@H]1O[C@@H](O[C@@H]2C[C@@H]3O[C@](O)(C[C@@H](O)C[C@@H](O)[C@H](O)CC[C@@H](O)C[C@@H](O)CC(=O)O[C@@H](C)[C@H](C)[C@H](O)[C@@H](C)\C=C\C=C\C=C\C=C\C=C\C=C\C=C\2)C[C@H](O)[C@H]3C(O)=O)[C@@H](O)[C@@H](N)[C@@H]1O

InChI

1S/C47H73NO17/c1-27-17-15-13-11-9-7-5-6-8-10-12-14-16-18-34(64-46-44(58)41(48)43(57)30(4)63-46)24-38-40(45(59)60)37(54)26-47(61,65-38)25-33(51)22-36(53)35(52)20-19-31(49)21-32(50)23-39(55)62-29(3)28(2)42(27)56/h5-18,27-38,40-44,46,49-54,56-58,61H,19-26,48H2,1-4H3,(H,59,60)/b6-5+,9-7+,10-8+,13-11+,14-12+,17-15+,18-16+/t27-,28-,29-,30+,31+,32+,33-,34-,35+,36+,37-,38-,40+,41-,42+,43+,44-,46-,47+/m0/s1

InChI key

APKFDSVGJQXUKY-INPOYWNPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Amphotericin B is a polyene antifungal antibiotic from Streptomyces sp. and has a high affinity for sterols, especially ergosterols, of both fungal and bacterial cell membranes.

Application

Amphotericin B is used to study the affects of the influx of calcium on antifungal activity, [1] and to study the binding and kinetics of Amphotericin B to sterols in monolayers, aqueous solutions, and phospholipid bilayers [2][3]. It is also commonly used in tissue culture as an antifungal supplement.
Amphotericin B from Streptomyces sp. has been used-
  • for amphotericin perforated patch technique[4]
  • as a reference antifungal drug during MIC (minimum inhibitory concentration) agar dilution assay[5][6][7]
  • as a component of Arcobacter selective isolation broth[8]

Biochem/physiol Actions

Amphotericin B binds to sterols, forming pores in the membrane, and causing small molecules to leak out. The product is most effective against fungi and yeast.
Mode of Action: Amphotericin B binds to sterols, forming pores in the membrane, and causing small molecules to leak out.

Antimicrobial spectrum: Includes fungi and yeast

Features and Benefits

High quality antibiotic suitable for mulitple research applications

Preparation Note

Amphotericin B is insoluble in water at pH 6 to 7, but soluble in water at pH 2 or 11. It is soluble in DMSO (30-40 mg/mL) and in dimethylformamide (2-4 mg/mL). Aqueous solutions cannot be sterile filtered due to poor solubility.

Amphotericin B remains active for 3 days in culture at 37°C. At -20°C, and with protection from air and light, the product remains active for 5 years.

Storage and Stability

Tightly closed. Dry. Keep in a well -ventilated place. Keep locked up or in an area accessible

Other Notes

For additional information on our range of Biochemicals, please complete this form.
Keep container tightly closed in a dry and well-ventilated place. Product is sensitive to light and moisture. Keep in a dry place.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maki Onda et al.
Journal of biochemistry, 134(1), 121-128 (2003-08-29)
To investigate the susceptibilities of fungal and mammalian cells to amphotericin B (AmB), AmB-loaded lysophosphatidylcholine (LPC)micelles as drug delivery vehicles were incubated at 37 degrees C with phosphatidylcholine vesicles containing different sterols as model systems for fungal and mammalian cells.
P David Rogers et al.
The Journal of antimicrobial chemotherapy, 51(2), 305-312 (2003-02-04)
The ability of amphotericin B to increase intracellular calcium concentrations in human cells is associated with the toxicity of this antifungal agent. The present study was performed to determine whether amphotericin B affects the influx or efflux of calcium in
Screening of antimicrobial activity of essential oil and methanol extract of Satureja hortensis on foodborne bacteria and fungi.
Adiguzel A, et al.
Czech Journal of Food Sciences, 25, 81-81 (2007)
Hakan Ozer et al.
Journal of agricultural and food chemistry, 55(3), 937-942 (2007-02-01)
Hippomarathrum microcarpum grows wild in eastern Anatolia, Turkey, and is a plant utilized as food by people. In this study, the in vitro antimicrobial and antioxidant activities of the essential oil and methanol extract from H. microcarpum and its essential
Biological activities of the essential oils and methanol extract of Origanum vulgare ssp. vulgare in the Eastern Anatolia region of Turkey.
Sahin F, et al.
Food Control, 15, 549-557 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service