Skip to Content
Merck
All Photos(1)

Key Documents

95209

Sigma-Aldrich

L-Ascorbic acid

BioUltra, ≥99.5% (RT)

Synonym(s):

L-Threoascorbic acid, Antiscorbutic factor, Vitamin C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O6
CAS Number:
Molecular Weight:
176.12
Beilstein:
84272
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.79
Pricing and availability is not currently available.

biological source

synthetic

grade

biotech. grade

product line

BioUltra

Assay

≥99.5% (RT)

form

powder or crystals

impurities

insoluble matter, passes filter test

ign. residue

≤0.05% (as SO4)

loss

≤0.05% loss on drying, 20 °C (HV)

color

white to off-white

pH

1.0-2.5 (25 °C, 1 M in H2O)

mp

190-194 °C (dec.)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤20 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤25 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

absorption

cut-off at 306 nm in H2O at 1 M

SMILES string

OC([C@]([C@@H](O)CO)([H])O1)=C(O)C1=O

InChI

1S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2,5,7-10H,1H2/t2-,5+/m0/s1

InChI key

CIWBSHSKHKDKBQ-JLAZNSOCSA-N

Gene Information

human ... SLC23A2(9962)

Looking for similar products? Visit Product Comparison Guide

General description

L-Ascorbic acid, also known as vitamin C, is a water-soluble vitamin.[1] It is naturally present in a variety of foods such as citrus fruits and vegetables. It is present only in minimal amount in milk.[2]

Application

L-Ascorbic acid has been used as an antioxidant supplement in incubation solution for dissected and sliced brain sections.[3][4] It has also been used as a supplement in phosphate-buffered saline (PBS) wash buffer during oxidative GFP chromophore formation.[5]

Biochem/physiol Actions

L-Ascorbic acid (Vitamin C) acts as a potential anti-oxidant[6] and reducing agent.[7] It prevents bacterial mutagenesis induced by N-nitroso compounds.[8] In plants, vitamin C elicits several cellular processes such as growth, apoptosis, pathogen responses, hormone responses, flowering and senescence, and protection against environmental stresses. [7] Vitamin C deficiency is associated with the development of scurvy, prominent - gingivitis, perifollicular hemorrhages, poor wound healing, petechiae and ecchymosis.[2]

Other Notes

Denaturation of proteinscitation; Dimeric association of E. coli RNA polymerase α subunits, studied by cleavage of single cysteine α subunits conjugated to Fe-BABEcitation.
Denaturation of proteins[9]; Dimeric association of E. coli RNA polymerase α subunits, studied by cleavage of single cysteine α subunits conjugated to Fe-BABE[10].

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Calcium Imaging Techniques In Vitro to Explore the Role of Dendrites in Signaling Physiological Action Potential Patterns
Neuromethods, 165-187 (2012)
Calcium Imaging Techniques In Vitro to Explore the Role of Dendrites in Signaling Physiological Action Potential Patterns
Bonnan A, et al.
The Journal of applied nutrition (1953)
F C Brown et al.
Neurochemical research, 3(3), 357-366 (1978-06-01)
Under conditions that are optimum for DbetaH, ascorbic acid denatures serum albumin, gamma-globulin, catalase, and DbetaH. With ascrobate plus Cu2+, the proteins are almost completely destroyed. Pyrazole protects DbetaH and albumin, but not catalase. Superoxide dismutase (SOD) is not denatured
Inhibition by L-ascorbate of bacterial mutagenesis induced by two N-nitroso compounds
Guttenplan JB
Nature, 268(5618), 368-368 (1977)
L-Ascorbate biosynthesis in higher plants: the role of VTC2
Linster CL and Clarke SG
Trends in Plant Science, 13(11), 567-573 (2008)

Questions

  1. The material is in powder or crystal form and whether it’s suitable for hydrogen evolution reaction (HER) measurements?

    1 answer
    1. The material specification is 'powder or crystals', which means it can vary from batch to batch. Ascorbic acid can donate electrons, facilitating the reduction of protons (H?) to hydrogen gas (H?). Its ability to undergo oxidation makes it a suitable candidate for these hydrogen evolution reactions. This product has not been tested internally for this application. Given the high purity of this material, it is likely suitable, but this would need to be determined by the end user.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service