Skip to Content
Merck
All Photos(4)

Key Documents

255564

Sigma-Aldrich

L-Ascorbic acid

ACS reagent, ≥99%

Synonym(s):

L-Threoascorbic acid, Antiscorbutic factor, Vitamin C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O6
CAS Number:
Molecular Weight:
176.12
Beilstein:
84272
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Assay

≥99%

form

crystals or granules
powder or chunks

optical activity

[α]25/D +20.5 to +21.5° in H2O

technique(s)

HPLC: suitable
cell culture | mammalian: suitable

ign. residue

<0.1%

color

white

pH

1.0-2.5 (25 °C, 176 g/L in water)

mp

190-194 °C (dec.)

solubility

water: soluble 176 g/L at 20 °C

cation traces

Fe: ≤0.001%
heavy metals: ≤0.002% (by ICP-OES)

application(s)

cell analysis

functional group

ester
hydroxyl
ketone

SMILES string

OC([C@]([C@@H](O)CO)([H])O1)=C(O)C1=O

InChI

1S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2,5,7-10H,1H2/t2-,5+/m0/s1

InChI key

CIWBSHSKHKDKBQ-JLAZNSOCSA-N

Gene Information

human ... SLC23A2(9962)

Looking for similar products? Visit Product Comparison Guide

General description

L-Ascorbic acid or vitamin C is a water-soluble vitamin found in various foodstuffs. Benefits of vitamin C may include protection against immune system deficiencies, cardiovascular disease, hyperpigmentation, skin protection, anti-aging, iron absorption, and red blood cell production. It is also useful in the biosynthesis of collagen, L-carnitine, and neurotransmitters.[1][2][3][4]

Application

L-ascorbic acid (L-AA) can be used as a reducing as well as a capping agent in the synthesis of well-dispersed metal nanoparticles and also in the reduction of graphene oxide sheets.[5][6]
Its immunostimulant derivatives play an important role in cell regulation.[7]

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Acta Biochimica et Biophysica Hungarica, 22, 167-167 (1987)
Synthesis of highly stable dispersions of nanosized copper particles using L-ascorbic acid
Xiong J, et al.
Green Chemistry, 13(4), 900-904 (2011)
Vitamin C: One compound, several uses. Advances for delivery, efficiency and stability
Carita AC, et al.
Nanomedicine: Nanotechnology, Biology, and Medicine, 31(2), 102117-102117 (2019)
Vitamin c
Linster CL and Van SE
FEBS Journal, 274(1), 1-22 (2007)
A review of micronutrients in sepsis: the role of thiamine, l-carnitine, vitamin C, selenium and vitamin D
Belsky JB, et al.
Nutrition Research Reviews, 31(2), 281-290 (2018)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service