Skip to Content
Merck
All Photos(4)

Documents

795437

Sigma-Aldrich

L-Ascorbic acid

anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%

Synonym(s):

L-Threoascorbic acid, Antiscorbutic factor, Vitamin C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O6
CAS Number:
Molecular Weight:
176.12
Beilstein:
84272
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.21

biological source

synthetic (organic)

Quality Level

grade

ACS reagent
anhydrous

product line

Redi-Dri

Assay

≥99%

form

crystals or granules
powder or chunks

optical activity

[α]25/D +20.5 to +21.5° in water

quality

free-flowing

ign. residue

<0.1%

color

white

pH

1.0-2.5 (25 °C, 176 g/L in water)

mp

190-194 °C (dec.)

solubility

water: soluble 176 g/L at 20 °C

cation traces

Fe: ≤0.001%
heavy metals: ≤0.002% (by ICP-OES)

SMILES string

OC([C@]([C@@H](O)CO)([H])O1)=C(O)C1=O

InChI

1S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2,5,7-10H,1H2/t2-,5+/m0/s1

InChI key

CIWBSHSKHKDKBQ-JLAZNSOCSA-N

Gene Information

human ... SLC23A2(9962)

Looking for similar products? Visit Product Comparison Guide

General description

L-Ascorbic acid or vitamin C is a water-soluble vitamin found in various foodstuffs. Benefits of vitamin C may include protection against immune system deficiencies, cardiovascular disease, hyperpigmentation, skin protection, anti-aging, iron absorption, and red blood cell production. It is also useful in the biosynthesis of collagen, L-carnitine, and neurotransmitters.

Application

L-Ascorbic acid (anhydrous) can be used in pharmaceutical applications.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weihua Li et al.
Journal of orthopaedic surgery and research, 15(1), 143-143 (2020-04-16)
Evidence suggests that microRNAs (miRNAs) regulate the expression of genes involved in bone metabolism. This study aimed to investigate the role of miR-505 in the osteogenic differentiation of MC3T3-E1 cells. We performed miRNA sequencing to identify differentially expressed miRNAs between
Vitamin C: One compound, several uses. Advances for delivery, efficiency and stability
Carita AC, et al.
Nanomedicine: Nanotechnology, Biology, and Medicine, 31(2), 102117-102117 (2019)
Juan Xiang et al.
Molecular medicine reports, 23(4) (2021-02-13)
Human periodontal ligament stem cells (hPDLSCs) associated with bone regeneration serve an important role in the treatment of periodontal disease. Long non‑coding RNAs are involved in the osteogenesis of multiple stem cells and can act as a sponge of microRNAs
Vitamin c
Linster CL and Van SE
FEBS Journal, 274(1), 1-22 (2007)
A review of micronutrients in sepsis: the role of thiamine, l-carnitine, vitamin C, selenium and vitamin D
Belsky JB, et al.
Nutrition Research Reviews, 31(2), 281-290 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service