Skip to Content
Merck
All Photos(1)

Key Documents

V-038

Supelco

L-Ascorbic acid solution

L-Ascorbic acid solution

1.0 mg/mL in acetonitrile: water, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile: water

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

shipped in

wet ice

storage temp.

−70°C

InChI

1S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2,5,7-10H,1H2/t2-,5+/m0/s1

InChI key

CIWBSHSKHKDKBQ-JLAZNSOCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-Ascorbic Acid (Vitamin C) is a water-soluble vitamin that is essential for growth and repair of tissue. Quantification of vitamin C levels is performed to evaluate vitamin C deficiency.

Application

L-Ascorbic acid solution (Vitamin C) may be used as a certified reference material (CRM) for the determination of the analyte in pharmaceutical formulations,[1] fruits and vegetables,[2] wines[3] and biological samples[3] by chromatography and spectrophotometry techniques.
This reference solution is suitable for use as starting material in the preparation of linearity standards, calibrators, or controls for identification of L-ascorbic acid in clinical diagnostic testing of biological samples or quality control testing of vitamin C foods or dietary supplements using HPLC with ultraviolet (UV) or electrochemical (EC) detection.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

75.2 °F

Flash Point(C)

24 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of l-ascorbic acid in human serum by chemiluminescence based on hydrogen peroxide--sodium hydrogen carbonate--CdSe/CdS quantum dots system
Chen H, et al.
Talanta, 81(4-5), 1688-1696 (2010)
Determination of l-ascorbic acid in wines by direct injection liquid chromatography using a polymeric column
Lopes P, et al.
Analytica Chimica Acta, 555(2), 242-245 (2006)
Flow injection spectrophotometric determination of L-ascorbic acid in pharmaceutical formulations with on-line solid-phase reactor containing copper (II) phosphate
Pereira AV and Fatibello-Filho O
Analytica Chimica Acta, 366(1-3), 55-62 (1998)
A rapid method for the determination of total L-ascorbic acid in fruits and vegetables by micellar electrokinetic capillary chromatography
Thompson CO and Trenerry VC
Food Chemistry, 53(1), 43-50 (1995)

Questions

Reviews

Active Filters

    • Reviews 2
    • Votes 0
    2 out of 5 stars.

    Lower concentration of ascorbic acid

    I wish I could get high concentration one. But they didn't have it in the solution

    Helpful?

    1. Response from MilliporeSigma:

      Thank you for your review. We are sorry to hear that your experience did not match your product expectations. We encourage customers who experience problems with our products to call or email us for additional technical support. Please visit https://www.sigmaaldrich.com/US/en/support/customer-support to submit a Product Technical Inquiry.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service