Skip to Content
Merck
All Photos(1)

Documents

1.00468

Supelco

L(+)-Ascorbic Acid

for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

L(+)-Ascorbic Acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
176.12
MDL number:
UNSPSC Code:
12352106
EC Index Number:
200-066-2
NACRES:
NA.21

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

product line

EMSURE®

Assay

99.0-100.5% (iodometric)

form

crystalline powder

autoignition temp.

380 °C

potency

11900 mg/kg LD50, oral (Rat)

technique(s)

thin layer chromatography (TLC): suitable

impurities

≤0.1% Related substances (unspecified impurities singly) (HPLC)
≤0.15% Related substances (Impurity C) (HPLC)
≤0.15% Related substances (Impurity D) (HPLC)
≤0.2% Oxalic acid
≤0.2% Related substances (sum of impurities (except impurity A and D)) (HPLC)

sulfated ash (600 °C)

≤0.05%

loss

≤0.1% loss on drying, 105 °C

color

clear

pH

2.2-2.5 (20 °C, 50 g/L in H2O)

solubility

330 g/L

density

1.65 g/cm3 at 20 °C

bulk density

500‑900 kg/m3

anion traces

chloride (Cl-): ≤50 ppm
sulfate (SO42-): ≤20 ppm

cation traces

Cu: ≤5 ppm
Fe: ≤2 ppm
heavy metals: ≤10 ppm

storage temp.

15-25°C

InChI

1S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2,5,7-8,10-11H,1H2

InChI key

TYQCGQRIZGCHNB-UHFFFAOYSA-N

General description

for analysis EMSURE® ACS,Reag. Ph Eur

Application


  • Sensitive determination in metabolic research: A study utilized (+/-)-4-Hydroxy-3-methoxymandelic Acid-D₃ in a gas chromatography-negative-ion chemical-ionization mass spectrometry analysis, providing a method for sensitive determination of tyrosine metabolites. This application supports the use of deuterated compounds in stable isotope-labeled tracer experiments to investigate metabolism in humans (Shimamura et al., 1986).

Analysis Note

Assay (iodometric)99.0 - 100.5 %Identity (IR-spectrum)conformsAppearancewhite or almost white, cristalline powderAppearance of solution (50 g/l CO₂-free water)clear (≤ 3 NTU) and not so intense in colour than reference solution BY₇pH (50 g/l CO₂-free water)2.1 - 2.6Spec. rotation [α²0/D (100 g/l, water)+20.5 - +21.5 °Chloride (Cl)≤ 50 ppmSulfate (SO₄)≤ 20 ppmCu (Copper)≤ 5 ppmFe (Iron)≤ 2 ppmHeavy metals (ACS)≤ 10 ppmOxalic acid≤ 0.2Related substances (HPLC) (Impurity C)≤ 0.15 %Related substances (HPLC) (Impurity D)≤ 0.15 %Related substances (HPLC) (unspecified impurities singly)≤ 0.10 %Related substances (HPLC) (sum of impurities (except impurity A and D))≤ 0.2 %Sulfated ash (600 °C)≤ 0.05 %Loss on Drying (105 °C)≤ 0.1 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service