Skip to Content
Merck
All Photos(1)

Key Documents

5.43808

Supelco

Acetic acid

100%, for HPLC LiChropur

Synonym(s):

Acetic acid, Ethanoic acid, Glacial acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2H
CAS Number:
Molecular Weight:
60.05
Beilstein:
506007
MDL number:
UNSPSC Code:
51242002
EC Index Number:
200-580-7
NACRES:
NA.21
Pricing and availability is not currently available.

Product Name

Acetic acid 100%, for HPLC LiChropur

Agency

suitable for ASTM® 7968
suitable for ASTM® 7978
suitable for DIN 38407-42
suitable for EPA 1633
suitable for EPA 533
suitable for EPA 8327
suitable for EPA ACB B21-02
suitable for ISO 21675 2019
suitable for ISO 25101
suitable for ISO/CEN 15968-2010

Quality Level

vapor density

2.07 (vs air)

vapor pressure

15.4 hPa ( 20 °C)

Assay

≥99.8% (alkalimetric)

form

liquid

autoignition temp.

485 °C
800 °F

expl. lim.

16 %, 92 °F
4 %, 59 °F

technique(s)

HPLC: suitable

refractive index

n20/D 1.371 (lit.)

pH

2.5 (20 °C, 50 g/L in H2O)

kinematic viscosity

1.17 cSt(20 °C)

bp

117-118 °C (lit.)

mp

16.2 °C (lit.)

transition temp

flash point 39 °C

density

1.04 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.02 ppm
Ca: ≤10.0 ppm
Cu: ≤0.010 ppm
Fe: ≤0.050 ppm
K: ≤0.10 ppm
Mg: ≤0.050 ppm
NH4+: ≤10.0 ppm
Na: ≤0.20 ppm

SMILES string

[F2C(F2C)13F3C]C(O)=O

application(s)

PFAS testing

storage temp.

15-25°C

InChI

1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)

InChI key

QTBSBXVTEAMEQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aceticacid 100% is a high-purity chemical reagent used for high-performance liquidchromatography (HPLC) applications. It is used as a mobile phase modifier orsolvent additive to adjust the pH and enhance the separation of analytes.[1]

Application

Acetic acid 100% can be used to adjust the pH of the sodium heptane sulfonate solution for the subsequent analysis of aciclovir and ganciclovir in human plasma by liquid chromatography–spectrofluorimetric method.[1]

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

102.2 °F

Flash Point(C)

39 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of phenolic constituents of biological interest in red wines by high-performance liquid chromatography
Lopez M, et al.
Journal of Chromatography A, 922(1-2) (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service