Skip to Content
Merck
All Photos(3)

Key Documents

1.00063

Supelco

Acetic acid (glacial)

100%, anhydrous for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Synonym(s):

Acetic acid, Ethanoic acid, Glacial acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2H
CAS Number:
Molecular Weight:
60.05
Beilstein:
506007
MDL number:
UNSPSC Code:
12352106
EC Index Number:
200-580-7
NACRES:
NA.21

Product Name

Acetic acid (glacial) 100%, anhydrous for analysis EMSURE® ACS,ISO,Reag. Ph Eur

grade

ACS reagent

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

vapor density

2.07 (vs air)

vapor pressure

15.4 hPa ( 20 °C)

product line

EMSURE®

Assay

≥99.8% (alkalimetric)

form

liquid

autoignition temp.

485 °C
800 °F

potency

3310 mg/kg LD50, oral (Rat)

expl. lim.

16 %, 92 °F
4 %, 59 °F

technique(s)

UV/Vis spectroscopy: suitable

refractive index

n20/D 1.371 (lit.)

pH

2.5 (20 °C, 50 g/L in H2O)

kinematic viscosity

1.17 cSt(20 °C)

bp

117-118 °C (lit.)

mp

16.2 °C (lit.)

transition temp

flash point 39 °C

solubility

soluble 602.9 g/L

density

1.04 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.4 ppm
phosphate (PO43-): ≤0.4 ppm
sulfate (SO42-): ≤0.4 ppm

cation traces

Ag: ≤0.005 ppm
Al: ≤0.020 ppm
As: ≤0.010 ppm
Au: ≤0.010 ppm
B: ≤0.100 ppm
Ba: ≤0.010 ppm
Be: ≤0.005 ppm
Bi: ≤0.050 ppm
Ca: ≤0.100 ppm
Cd: ≤0.020 ppm
Co: ≤0.010 ppm
Cr: ≤0.020 ppm
Cu: ≤0.010 ppm
Fe: ≤0.050 ppm
Ga: ≤0.050 ppm
Ge: ≤0.020 ppm
Hg: ≤0.005 ppm
In: ≤0.050 ppm
K: ≤0.100 ppm
Li: ≤0.010 ppm
Mg: ≤0.050 ppm
Mn: ≤0.010 ppm
Mo: ≤0.010 ppm
Na: ≤0.200 ppm
Ni: ≤0.020 ppm
Pb: ≤0.010 ppm
Pt: ≤0.100 ppm
Sn: ≤0.050 ppm
Sr: ≤0.010 ppm
Ti: ≤0.050 ppm
Tl: ≤0.020 ppm
V: ≤0.010 ppm
Zn: ≤0.030 ppm
Zr: ≤0.050 ppm
heavy metals (as Pb): ≤0.5 ppm

SMILES string

[F2C(F2C)13F3C]C(O)=O

storage temp.

15-25°C

InChI

1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)

InChI key

QTBSBXVTEAMEQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Acetic acid (glacial) 100% (EMSURE® grade product) is optimized for regulated analyses and highly demanding lab applications. It offers the highest quality and unmatched scope of specifications to give complete control of test conditions and eliminate uncertainties. This reagent is ideal for demanding analytical procedures that need low detection limits and maximum sensitivity.

Features and Benefits

Advantages of this product:      
  • Streamlined lab work      
  • Time and cost savings      
  • Superior comparability of results      
  • Certainty during product use      
  • Accuracy regarding impurities      
  • Confidence in analysis and production

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

102.2 °F

Flash Point(C)

39 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service