1.00062
Acetic acid
96%, for analysis EMSURE®
Synonym(s):
Acetic acid solution, Ethanoic acid
About This Item
product name
Acetic acid 96%, for analysis EMSURE®
vapor pressure
15.2 hPa ( 20 °C)
Quality Level
product line
EMSURE®
Assay
≥96.0% (alkalimetric)
form
liquid
autoignition temp.
463 °C
expl. lim.
4-19.9 % (v/v)
impurities
≤0.01 ppm As (Arsenic)
≤0.010 ppm Ag (Silver)
≤0.010 ppm Au (Gold)
≤0.010 ppm Ba (Barium)
≤0.010 ppm Co (Cobalt)
≤0.010 ppm Li (Lithium)
≤0.010 ppm Mn (Manganese)
≤0.010 ppm Sr (Strontium)
≤0.010 ppm V (Vanadium)
≤0.020 ppm Be (Beryllium)
≤0.020 ppm Cr (Chromium)
≤0.020 ppm Cu (Copper)
≤0.020 ppm Mo (Molybdenum)
≤0.020 ppm Ni (Nickel)
≤0.020 ppm Pb (Lead)
≤0.050 ppm Al (Aluminium)
≤0.050 ppm Cd (Cadmium)
≤0.050 ppm Ga (Gallium)
≤0.050 ppm Ge (Germanium)
≤0.050 ppm In (Indium)
≤0.050 ppm Mg (Magnesium)
≤0.050 ppm Sn (Tin)
≤0.050 ppm Tl (Thallium)
≤0.050 ppm Zn (Zinc)
≤0.100 ppm B (Boron)
≤0.100 ppm Bi (Bismuth)
≤0.100 ppm Fe (Iron)
≤0.100 ppm K (Potassium)
≤0.100 ppm Pt (Platinum)
≤0.100 ppm Ti (Titanium)
≤0.100 ppm Zr (Zirconium)
≤0.200 ppm Ca (Calcium)
≤0.5 ppm Chloride (Cl)
≤0.5 ppm Phosphate (PO4)
≤0.5 ppm Sulfate (SO4)
≤0.500 ppm Na (Sodium)
≤2 ppm Acetaldehyde
SMILES string
CC(O)=O
evapn. residue
≤5 ppm
color
APHA: ≤10
pH
2.5 (20 °C, 10 g/L in H2O)
bp
117.9 °C/1013 hPa
mp
16.6 °C
transition temp
flash point 39 °C
density
1.05 g/cm3 at 20 °C
storage temp.
15-25°C
InChI
1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)
InChI key
QTBSBXVTEAMEQO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Analysis Note
Color: ≤ 10 Hazen
Acetaldehyde: ≤ 2 ppm
Chloride (Cl): ≤ 0.5 ppm
Phosphate (PO₄): ≤ 0.5 ppm
Sulfate (SO₄): ≤ 0.5 ppm
Ag (Silver): ≤ 0.010 ppm
Al (Aluminium): ≤ 0.050 ppm
As (Arsenic): ≤ 0.01 ppm
Au (Gold): ≤ 0.010 ppm
B (Boron): ≤ 0.100 ppm
Ba (Barium): ≤ 0.010 ppm
Be (Beryllium): ≤ 0.020 ppm
Bi (Bismuth): ≤ 0.100 ppm
Ca (Calcium): ≤ 0.200 ppm
Cd (Cadmium): ≤ 0.050 ppm
Co (Cobalt): ≤ 0.010 ppm
Cr (Chromium): ≤ 0.020 ppm
Cu (Copper): ≤ 0.020 ppm
Fe (Iron): ≤ 0.100 ppm
Ga (Gallium): ≤ 0.050 ppm
Ge (Germanium): ≤ 0.050 ppm
In (Indium): ≤ 0.050 ppm
K (Potassium): ≤ 0.100 ppm
Li (Lithium): ≤ 0.010 ppm
Mg (Magnesium): ≤ 0.050 ppm
Mn (Manganese): ≤ 0.010 ppm
Mo (Molybdenum): ≤ 0.020 ppm
Na (Sodium): ≤ 0.500 ppm
Ni (Nickel): ≤ 0.020 ppm
Pb (Lead): ≤ 0.020 ppm
Pt (Platinum): ≤ 0.100 ppm
Sn (Tin): ≤ 0.050 ppm
Sr (Strontium): ≤ 0.010 ppm
Ti (Titanium): ≤ 0.100 ppm
Tl (Thallium): ≤ 0.050 ppm
V (Vanadium): ≤ 0.010 ppm
Zn (Zinc): ≤ 0.050 ppm
Zr (Zirconium): ≤ 0.100 ppm
Indifference against chromic acid: conforms
Substances reducing potassium permanganate: conforms
Evaporation residue: ≤ 5 ppm
Dilution test: conforms
Date of expiry: see product label
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
102.2 °F
Flash Point(C)
39 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service