Skip to Content
Merck
All Photos(1)

Documents

361593

Sigma-Aldrich

Artemisinin

98%

Synonym(s):

Arteannuin, Qinghaosu

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22O5
CAS Number:
Molecular Weight:
282.33
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.79

Assay

98%

form

solid

optical activity

[α]20/D +76°, c = 0.5 in methanol

mp

156-157 °C (lit.)

antibiotic activity spectrum

parasites
viruses

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

Mode of action

enzyme | inhibits

SMILES string

C[C@@H]1CC[C@H]2[C@@H](C)C(=O)O[C@@H]3OC4(C)CC[C@@H]1[C@@]23OO4

InChI

1S/C15H22O5/c1-8-4-5-11-9(2)12(16)17-13-15(11)10(8)6-7-14(3,18-13)19-20-15/h8-11,13H,4-7H2,1-3H3/t8-,9-,10+,11+,13-,14-,15-/m1/s1

InChI key

BLUAFEHZUWYNDE-NNWCWBAJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Artemisinin is extracted from sweet wormwood (Artemisia annua). It has anticancer, antiprotozoal and antiviral activities. Artemisinin is associated with brainstem encephalopathy.

Application

Artemisinin has been used:
  • in drug susceptibility assays
  • as a standard for its quantification from artemisinin from hairy roots
  • to check cell viability using 3-(4, 5-dimethylthiazol-2yl)-2,5-diphenyl tetrazolium bromide (MTT) assay
  • as an antimalarial agent to investigate the role of autophagy-related (ATG) genes throughout the P. falciparum asexual and sexual blood stages

Biochem/physiol Actions

Artemisinin (Qinghaosu), a sesquiterpene lactone, is a highly active anti-malarial (falciparum malaria) drug. Artemisinin is also an anthelmintic (parasitic worm) effective against the blood fluke, schistosomiasis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pierre-Eric Campos et al.
Marine drugs, 17(3) (2019-03-17)
Chemical study of the CH₂Cl₂-MeOH (1:1) extract of the sponge Fascaplysinopsis reticulata collected in Mayotte highlighted three new tryptophan derived alkaloids, 6,6'-bis-(debromo)-gelliusine F (1), 6-bromo-8,1'-dihydro-isoplysin A (2) and 5,6-dibromo-8,1'-dihydro-isoplysin A (3), along with the synthetically known 8-oxo-tryptamine (4) and the
Stimulation of artemisinin synthesis by combined cerebroside and nitric oxide elicitation in Artemisia annua hairy roots
Wang JW, et al.
Applied Microbiology and Biotechnology, 85(2), 285-292 (2009)
Artemether resistance in vitro is linked to mutations in PfATP6 that also interact with mutations in PfMDR1 in travellers returning with Plasmodium falciparum infections
Pillai DR, et al.
Malaria Journal, 11(1), 131-131 (2012)
Olivo Miotto et al.
Nature genetics, 45(6), 648-655 (2013-04-30)
We describe an analysis of genome variation in 825 P. falciparum samples from Asia and Africa that identifies an unusual pattern of parasite population structure at the epicenter of artemisinin resistance in western Cambodia. Within this relatively small geographic area
The multifunctional autophagy pathway in the human malaria parasite, Plasmodium falciparum
Cervantes S, et al.
Autophagy, 10(1), 80-92 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service