Skip to Content
Merck
All Photos(1)

Key Documents

349356

Sigma-Aldrich

Platinum

foil, thickness 0.05 mm, 99.99% trace metals basis

Synonym(s):

Platinum black, Platinum element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pt
CAS Number:
Molecular Weight:
195.08
EC Number:
MDL number:
UNSPSC Code:
12141734
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99% trace metals basis

form

foil

resistivity

10.6 μΩ-cm, 20°C

thickness

0.05 mm

bp

3827 °C (lit.)

mp

1772 °C (lit.)

density

21.45 g/cm3 (lit.)

SMILES string

[Pt]

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

Related Categories

General description

Platinum foils may be used to fabricate multi-electrode spiral nerve cuffs. It was found to be the most active catalyst in the kinetic study of catalytic oxidation of alkanes over platinum foils.

Application

  • Platinum-group elements: This report covers the essential physical and chemical properties of platinum and its related metals, which are vital for applications ranging from automotive catalysts to anticancer drugs (Zientek et al., 2017).
  • Platinum-based drugs for cancer therapy and anti-tumor strategies: This review discusses the development and clinical use of platinum-based chemotherapy agents, highlighting new platinum compounds and their mechanisms of action, crucial for drug discovery and medicinal chemistry (Zhang et al., 2022).
  • Mastering the surface strain of platinum catalysts for efficient electrocatalysis: This article presents research on optimizing the surface properties of platinum for enhanced catalytic efficiency, which is significant for developing new catalytic materials in material science and energy applications (He et al., 2021).

Quantity

600 mg = (approx.) 25 mm × 25 mm

2.4 g = approximately 50 mm × 50 mm

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure characterization of platinum foil for neural stimulating electrodes
Peclin P, et al.
Bio-medical materials and engineering, 24, 1827-1835 (2014)
Kinetic study of the catalytic oxidation of alkanes over nickel, palladiium and platinum foils
Aryafar M and Zaera F
Catalysis Letters, 48, 173-183 (1997)
Yuan Xiong et al.
Organic letters, 15(8), 1962-1965 (2013-04-12)
Metal-mediated rearrangements of 3-alkynyl flavone ethers are reported. The overall process involves 5-endo enyne cyclization to a platinum-containing spiro-oxocarbenium intermediate which may be trapped with methanol to produce spirodihydrofurans or further rearranged to afford either allenyl chromanediones or benzofuranones.
Shai Rozen et al.
Plastic and reconstructive surgery, 131(6), 1253-1265 (2013-05-30)
Left untreated, paralytic lagophthalmos may result in corneal dryness, ulcerations, and subsequent blindness. The most common nondynamic surgical solution is upper eyelid weight placement in a superficial, pretarsal pocket, carrying the risk of visibility, extrusion, and entropion. The authors present
Loris De Cecco et al.
PloS one, 8(3), e58849-e58849 (2013-03-14)
We recently reported that peritumoral CpG-ODN treatment, activating TLR-9 expressing cells in tumor microenvironment, induces modulation of genes involved in DNA repair and sensitizes cancer cells to DNA-damaging cisplatin treatment. Here, we investigated whether this treatment induces modulation of miRNAs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service