Skip to Content
Merck
All Photos(1)

Key Documents

C7874

Sigma-Aldrich

Piperaquine tetraphosphate tetrahydrate

≥98% (HPLC)

Synonym(s):

7-Chloro-4-[4-[3-[4-(7-chloroquinolin-4-yl)piperazin-1-yl]propyl]piperazin-1-yl]quinoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H32Cl2N6·4H3PO4·4H2O
CAS Number:
Molecular Weight:
999.55
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to off-white

solubility

H2O: ≥10 mg/mL

storage temp.

room temp

SMILES string

O.O.O.O.OP(O)(O)=O.OP(O)(O)=O.OP(O)(O)=O.OP(O)(O)=O.Clc1ccc2c(ccnc2c1)N3CCN(CCCN4CCN(CC4)c5ccnc6cc(Cl)ccc56)CC3

InChI

1S/C29H32Cl2N6.4H3O4P.4H2O/c30-22-2-4-24-26(20-22)32-8-6-28(24)36-16-12-34(13-17-36)10-1-11-35-14-18-37(19-15-35)29-7-9-33-27-21-23(31)3-5-25(27)29;4*1-5(2,3)4;;;;/h2-9,20-21H,1,10-19H2;4*(H3,1,2,3,4);4*1H2

InChI key

AMCQDGFOKTXHSY-UHFFFAOYSA-N

Related Categories

Application

Piperaquine tetraphosphate tetrahydrate has been used to study its effect on the tert-butylhydroperoxide (t-BHP)-induced alterations of cell morphology and viability

Biochem/physiol Actions

Piperaquine tetraphosphate tetrahydrate is an antimalarial agent highly active against chloroquine-resistant Plasmodium falciparum and Plasmodium vivax.
Piperaquine tetraphosphate tetrahydrate is an autophagy inhibitor.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chuanhong Wu et al.
Scientific reports, 8(1), 574-574 (2018-01-14)
Neurodegenerative diseases (NDD) are typically associated with neuron loss in nervous system areas. Interventions with related death mechanisms may ameliorate NDD progression. Oxidative stress plays an important role in NDD cell death routines. However, tert-butylhydroperoxide (t-BHP), a widely used oxidative
Yingshan Han et al.
Journal of medical virology, 91(7), 1182-1190 (2019-02-26)
Studies aimed at repurposing existing drugs revealed that some antimalarial compounds possess anti-Zika virus (anti-ZIKV) activity. Here, we further tested 14 additional antimalarial drugs and their metabolites or analogs for anti-ZIKV activity using a phenotypic screening approach. We identified four
Chuanhong Wu et al.
Scientific reports, 8(1), 574-574 (2018-01-14)
Neurodegenerative diseases (NDD) are typically associated with neuron loss in nervous system areas. Interventions with related death mechanisms may ameliorate NDD progression. Oxidative stress plays an important role in NDD cell death routines. However, tert-butylhydroperoxide (t-BHP), a widely used oxidative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service