Skip to Content
Merck
All Photos(4)

Documents

57330-U

Supelco

SPME Fiber Holder

greener alternative

for use with manual sampling

Synonym(s):

Solid Phase Microextraction Fiber Holder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115724
NACRES:
NB.21

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

solid phase microextraction (SPME): suitable

compatibility

for use with manual sampling

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This is a Solid Phase Microextraction (SPME) technology product. SPME is one of the greener technologies for sample preparation as it is solvent free extraction technology and SPME fibres are re-usable and generates as little waste as possible, thus aligns with "Waste Prevention”, and ″Safer Solvents and Auxiliaries″. Click here for more information.
The holder protects the coated fiber, and controls exposure of the fiber during analyte adsorption and desorption. The holder is reusable indefinitely and accepts the replaceable fiber assembly. First time users must order both a holder and a fiber assembly.

Fiber Holder for Manual Sampling

An adjustable depth guide positions the fiber for sampling and for correct placement in the heated zone of the GC injection port. The fiber can be locked in the exposed position.

Fiber Holder for Automated Sampling or HPLC Analysis

Use this fiber holder with a Varian 8100/8200 AutoSampler or with our SPME/HPLC interface. An SPME upgrade kit is necessary for operation with the Varian AutoSampler - contact Varian Instrument Division for information concerning system requirements.

Fiber Holder for CTC CombiPAL/CombiPAL-xt and Gerstel MPS2

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edited by Sue Ann Scheppers Wercinski
Solid Phase Microextraction: A Practical Guide, 257-257 (1999)
Na Li et al.
Food chemistry, 332, 127426-127426 (2020-07-04)
A selected Pichia fermentans strain was simultaneously and sequentially inoculated in synthetic and real juice with S. cerevisiae strains of different antagonistic activities in a ratio 1:1 to observe the correlation between varietal odorants and glycosidase activities. Fermentations using pure
Pawliszyn, Janusz
Solid Phase Microextraction: Theory and Practice, 264-264 (1997)
Abdelhamid Neggad et al.
Chemistry & biodiversity, 16(10), e1900267-e1900267 (2019-08-17)
The volatile composition of seven honey samples collected from various regions of Algeria and feeding on different plants have been determined. The Headspace Solid-Phase MicroExtraction (HS-SPME) coupled with Gas Chromatography-Mass Spectrometry (GC/MS) was used to achieve the purpose. In this
Pawliszyn, Janusz
Applications of Solid Phase Microextraction, 674-674 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service