Skip to Content
Merck
All Photos(1)

Key Documents

L5420

Sigma-Aldrich

Lumefantrine

Synonym(s):

(9Z)-2,7-Dichloro-9-[(4-chlorophenyl)methylene]-α-[(dibutylamino)methyl]-9H-fluorene-4-methanol, Benflumetol, CPG-56695

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H32Cl3NO
CAS Number:
Molecular Weight:
528.94
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

Quality Level

form

powder

color

yellow

solubility

DMSO: 2 mg/mL, clear (warmed)

originator

Novartis

storage temp.

room temp

SMILES string

CCCCN(CCCC)CC(O)c1cc(Cl)cc2C(=C/c3ccc(Cl)cc3)\c4cc(Cl)ccc4-c12

InChI

1S/C30H32Cl3NO/c1-3-5-13-34(14-6-4-2)19-29(35)28-18-23(33)17-27-25(15-20-7-9-21(31)10-8-20)26-16-22(32)11-12-24(26)30(27)28/h7-12,15-18,29,35H,3-6,13-14,19H2,1-2H3/b25-15-

InChI key

DYLGFOYVTXJFJP-MYYYXRDXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lumefantrine, also known as benflumetol is an aryl-amino alcohol.[1] It is a member of the group of arylamine alcohols like quinine, mefloquine and halofantrine.[2]

Application

Lumefantrine has been used:
to study its effect on ex-vivo Plasmodium falciparum sensitivity using the tritiated hypoxanthine-based assay[3]
as a standard in the quantification of combined tablet formulation using HPTLC[4]
as a drug molecule in in vitro growth inhibition assay for in vitro B. caballi growth inhibition studies[5]

Biochem/physiol Actions

Lumefantrine is is an antimalarial for the treatment of multi-drug resistant strains of falciparum malaria.

Features and Benefits

This compound was developed by Novartis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reduced ex vivo susceptibility of Plasmodium falciparum after oral artemether-lumefantrine treatment in Mali
Dama S, et al.
Malaria Journal, 16(1), 59-59 (2017)
Thomas T Thomsen et al.
The American journal of tropical medicine and hygiene, 85(6), 979-983 (2011-12-07)
Tanzania implemented artemether-lumefantrine (AL) as the first-line treatment for uncomplicated malaria in November of 2006 because of resistance to sulfadoxine-pyrimethamine. AL remains highly efficacious, but widespread use may soon facilitate emergence of artemisinin tolerance/resistance, which initially may be detected at
Use of a model procedure for transfer of Minilab qualitative screening TLC methods for lumefantrine and artemether in a combined tablet formulation to individual and simultaneous quantitative HPTLC-densitometry methods
Nguyen M and Sherma J
Malaria Journal (2013)
Lumefantrine and o-choline-Parasite metabolism specific drug molecules inhibited in vitro growth of Theileria equi and Babesia caballi in MASP culture system
Maji C, et al.
Ticks and Tick-Borne Diseases, 10(3), 568-574 (2019)
Anti-infective agents
Drugs During Pregnancy and Lactation, 115-176 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service