Skip to Content
Merck
All Photos(4)

Documents

271748

Sigma-Aldrich

Pyridoxal hydrochloride

99%

Synonym(s):

3-Hydroxy-5-(hydroxymethyl)-2-methyl-4-pyridinecarboxaldehyde hydrochloride, PL HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9NO3 · HCl
CAS Number:
Molecular Weight:
203.62
Beilstein:
3656994
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.79

biological source

synthetic (Organic)

Assay

99%

form

powder

color

white to off-white

mp

173 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

Cl[H].[H]C(=O)c1c(CO)cnc(C)c1O

InChI

1S/C8H9NO3.ClH/c1-5-8(12)7(4-11)6(3-10)2-9-5;/h2,4,10,12H,3H2,1H3;1H

InChI key

FCHXJFJNDJXENQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pyridoxal, a form of vitamin B6, is a water soluble vitamin. The food sources of vitamin B6 include grains, nuts, fruits, vegetables and meat.

Application

Pyridoxal hydrochloride has been used to test its potential contribution to plant stress tolerance. It has also been used as a supplement in modified M9 minimal medium to culture E. coli BL21(DE3) cells for expression of 15N-labeled ATG101.
For the labeling of amino acids and their detection in picomolar amounts; Coenzymes and Cofactors, vol. 1: Vitamin B6, Pyridoxal Phosphate

Biochem/physiol Actions

Vitamin B6 plays a key role in metabolism of both carbohydrates and amino acids. It is implicated in the synthesis of neurotransmitters like dopamine from L-DOPA, serotonin from 5-HTP, and γaminobutyric acid (GABA) from glutamate. Vitamin B6 plays a major role in maintenance of a healthy nervous system.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The mammalian autophagy initiator complex contains 2 HORMA domain proteins
Michel M, et al.
Autophagy, 11(12), 2300-2308 (2015)
Hydrosoluble vitamins
Handbook of Clinical Neurology (2014)
Root metabolic plasticity underlies functional diversity in mycorrhiza-enhanced stress tolerance in tomato
Rivero J, et al.
The New phytologist, 220(4), 1322-1336 (2018)
B Vitamins in the nervous system: Current knowledge of the biochemical modes of action and synergies of thiamine, pyridoxine, and cobalamin
Calderon-Ospina CA, et al.
CNS Neuroscience & Therapeutics (2019)
Toshio Iwasaki et al.
Journal of the American Chemical Society, 131(38), 13659-13667 (2009-09-10)
CW EPR spectra of reduced [2Fe-2S](Cys)(3)(His)(1) clusters of mammalian mitoNEET soluble domain appear to produce features resulting from the interaction of the electron spins of the two adjacent clusters, which can be explained by employing the local spin model. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service