Skip to Content
Merck
All Photos(1)

Key Documents

47862

Supelco

Pyridoxine Hydrochloride (B6)

analytical standard

Synonym(s):

Pyridoxine hydrochloride, Adermine hydrochloride, PN HCl, Pyridoxol hydrochloride, Vitamin B6 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11NO3 · HCl
CAS Number:
Molecular Weight:
205.64
Beilstein:
3632435
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

feature

standard type water soluble vitamin

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

214-215 °C (lit.)

application(s)

pharmaceutical (small molecule)
vitamins, nutraceuticals, and natural products

format

neat

storage temp.

2-30°C

SMILES string

Cl[H].Cc1ncc(CO)c(CO)c1O

InChI

1S/C8H11NO3.ClH/c1-5-8(12)7(4-11)6(3-10)2-9-5;/h2,10-12H,3-4H2,1H3;1H

InChI key

ZUFQODAHGAHPFQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pyridoxine Hydrochloride (B6) is a white, odorless and crystalline powder type of vitamin. It can be used in the production of both dry and liquid type of pharmaceutical preparations. It can also be widely used in vitamin nutrification of foods. It is involved as a coenzyme vitamin in a variety of metabolic reactions and also shows therapeutic applications in the treatment of premenstrual and carpal tunnel syndrome, hyperemesis gravidarum in pregnancy, autism, etc.

Application

Pyridoxine hydrochloride may be used as an analytical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography and spectrophotometric techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hechtman L
Clinical naturopathic medicine (2013)
Pyridoxine Hydrochloride
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 3451-3452 (2017)
Spectrophotometric methods for the simultaneous analysis of meclezine hydrochloride and pyridoxine hydrochloride in bulk drug and pharmaceutical formulations
Arayne MS, et al.
Pakistan Journal of Pharmaceutical Sciences, 20(2), 149-156 (2007)
National Research Council (U.S.). Committee on Dietary Allowances
Human Vitamin B6 Requirements: Proceedings of a Workshop : Letterman Army Institute of Research, Presidio of San Francisco, California, June 11-12, 1976 (1978)
Simultaneous spectrophotometric analysis of a ternary mixture of pharmaceuticals?assay for meclozine hydrochloride, pyridoxine hydrochloride and caffeine
Sharma SC, et al.
Journal of Pharmaceutical and Biomedical Analysis, 7(3), 321-327 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service