Skip to Content
Merck
All Photos(1)

Documents

T8637

Sigma-Aldrich

Thiamine monophosphate chloride dihydrate

Synonym(s):

Aneurine monophosphate chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18ClN4O4PS · 2H2O
CAS Number:
Molecular Weight:
416.82
Beilstein:
3844977
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.79

biological source

synthetic

Assay

≥99.0% (HPLC)

form

powder

technique(s)

HPLC: suitable

color

white to off-white

mp

190 °C

storage temp.

2-8°C

SMILES string

[Cl-].Cc1ncc(C[n+]2csc(CCOP(O)(O)=O)c2C)c(N)n1

InChI

1S/C12H17N4O4PS.ClH/c1-8-11(3-4-20-21(17,18)19)22-7-16(8)6-10-5-14-9(2)15-12(10)13;/h5,7H,3-4,6H2,1-2H3,(H3-,13,14,15,17,18,19);1H

InChI key

GUGWNSHJDUEHNJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Thiamine monophosphate chloride dihydrate has been used as a standard in high-performance liquid chromatography (HPLC) to quantify thiamine in plant exudates and to evaluate its purity.

Biochem/physiol Actions

Thiamine monophosphate (TMP) is used as a substrate of thiamine-phosphate kinase (EC 2.7.4.16) and a precursor of thiamine pyrophosphate (TPP). TPP is a coenzyme involved in the catabolism of sugars and amino acids.
Thiamine monophosphate (TMP) is used as a substrate of thiamine-phosphate kinase (EC 2.7.4.16) and a precursor of thiamine pyrophosphate (TPP). TPP is a coenzyme involved in the catabolism of sugars and amino acids. TMP acts as an intermediate in the hydrolysis of thiamine pyrophosphate to thiamine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural studies of thiamin monophosphate kinase in complex with substrates and products
McCulloch KM, et al.
Biochemistry, 47(12), 3810-3821 (2008)
Maria Rapala-Kozik et al.
Plant physiology and biochemistry : PPB, 47(4), 237-242 (2009-01-27)
The pool of thiamine diphosphate (TDP), available for TDP-dependent enzymes involved in the major carbohydrate metabolic pathways, is controlled by two enzyme systems that act in the opposite directions. The thiamine pyrophosphokinase (TPK) activates thiamine into TDP and the numerous
Aminoff's Neurology and General Medicine, 47(12) (2014)
Purity of standard solutions of thiamine and its phosphate esters for HPLC analysis
Ihara H, et al.
Journal of Analytical Bio-Science, 25(3) (2002)
New Research and Developments of Water-Soluble Vitamins (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service