Skip to Content
Merck
All Photos(1)

Documents

82870

Sigma-Aldrich

Pyridoxal 5′-phosphate monohydrate

≥97.0% (NT)

Synonym(s):

P5P, 3-Hydroxy-2-methyl-5-([phosphonooxy]methyl)-4-pyridinecarboxaldehyde, Codecarboxylase, PLP, Pyridoxal 5-phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10NO6P · H2O
CAS Number:
Molecular Weight:
265.16
Beilstein:
234749
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic

Assay

≥97.0% (NT)

form

powder

technique(s)

enzyme immunoassay: suitable

color

white to light yellow

mp

140-143 °C (lit.)

anion traces

chloride (Cl-): ≤500 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤20 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤50 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤2000 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

application(s)

cell analysis

storage temp.

2-8°C

SMILES string

O=CC1=C(C(C)=NC=C1COP(O)(O)=O)O.[OH]

InChI

1S/C8H10NO6P.H2O/c1-5-8(11)7(3-10)6(2-9-5)4-15-16(12,13)14;/h2-3,11H,4H2,1H3,(H2,12,13,14);1H2

InChI key

CEEQUQSGVRRXQI-UHFFFAOYSA-N

General description

Pyridoxal 5′-phosphate (PLP), an active form of vitamin B6/ pyridoxine is the coenzyme of amino acid metabolism.

Application

Pyridoxal 5′-phosphate monohydrate has been used:
  • as a component in the reaction mixture for ornithine decarboxylase (ODC) activity assay
  • as a standard to quantify the concentration of pyridoxal 5′-phosphate (PLP) in cerebrospinal fluid (CSF) of children
  • as a dietary supplement to study its effects on the lethal phenotype of mutant flies

Biochem/physiol Actions

Pyridoxine participates in the production of lipid and neurotransmitters. Pyridoxal 5′-phosphate (PLP) serves as a cofactor to stabilize carbanions at Cα of amino acids. Pyridoxine deficiency results in peripheral neuropathy. Absence of PLP leads to neonatal epileptic encephalopathy. PLP acts as a coenzyme in transamination, decarboxylation and deamination reactions such as the conversion of dopa into dopamine; of glutamate into γ aminobutyric acid (GABA); of histidine to histamine. PLP works mechanistically through formation of a Schiff-base.

Other Notes

Coenzymes and Cofactors, vol. 1: Vitamin B6, Pyridoxal Phosphate

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regulation and function of pyridoxal phosphate in CNS
Metzler DE
Neurochemistry International, 3(3-4), 207-209 (1981)
Assay of Ornithine Decarboxylase and Spermidine/Spermine N1-acetyltransferase Activities
Hyvonen MT, et al.
Bio-protocol (2014)
Nutritional Deficiencies and Syndromes Associated with Alcoholism
Textbook of Clinical Neurology, 897-917 (2007)
Vitamin B6 biosynthesis: charting the mechanistic landscape.
Teresa B Fitzpatrick et al.
Chembiochem : a European journal of chemical biology, 11(9), 1185-1193 (2010-04-17)
Florence Bourquin et al.
Protein science : a publication of the Protein Society, 20(9), 1492-1508 (2011-06-29)
Sphingolipids are membrane constituents as well as signaling molecules involved in many essential cellular processes. Serine palmitoyltransferase (SPT) and sphingosine-1-phosphate lyase (SPL), both PLP (pyridoxal 5'-phosphate)-dependent enzymes, function as entry and exit gates of the sphingolipid metabolism. SPT catalyzes the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service