Skip to Content
Merck
All Photos(1)

Key Documents

14008C

Sigma-Aldrich

Serum Plus Medium Supplement

made with gamma irradiated FBS

Synonym(s):

FBS, FCS, GI FBS, GI FCS, GI Serum supplement, GI sera supplement

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

Quality Level

sterility

γ-irradiated by SER-TAIN process (FBS)
sterile; sterile-filtered

form

solution

shipped in

dry ice

storage temp.

−20°C

General description

SERUM PLUS contains a low level of gamma irradiated Fetal Bovine Serum (FBS) supplemented with proteins that promote nutrient transport, cell attachement, and cell growth.

Application

Serum Plus is a medium supplement that can be used as a serum substitute for most cell culture applications. It has been used as a supplement in media for maintenance of bloodstream form Trypanosoma brucei Lister 427.

Features and Benefits

Benefits include less than half of the total protein concentration found in serum, reduced levels of inhibitors such as proteases and viral antibodies, and consistency of lot-to-lot performance due to greater control of the component materials.

Preparation Note

Thawing
1. Remove the supplement bottles from the freezer and allow them to acclimate to room temperature for approximately 10 minutes.
2. Place each container in a 30 - 37 °C water bath or incubator. Excessive temperatures will degrade heat liable nutrients. If using a water bath, prevent the bottle caps from being completely submerged.
3. Gently swirl the bottles every 10-15 minutes until the serum is completely thawed.
4. After thawing, use the supplement promptly. Liquid supplement may be stored refrigerated (2 - 8 °C) up to four weeks. To avoid freeze/thaw cycles or long periods of refrigeration, it is recommended that any unused supplement be immediately dispensed into small, useful aliquots and refrozen for future use.

Note:
Gentle, periodic swirling of this product is crucial for optimum performance. If a bottle is not periodically swirled as it thaws, gradients containing high concentrations of salts, proteins and lipids will form throughout the liquid portion and lead to the formation of crystalline or flocculent precipitates. These cryoprecipitates are not toxic to cell cultures, but they affect the appearance and consistency of each bottle.

Small amounts of cryoprecipitates are not uncommon, and will not affect product performance. Gently warming and mixing the supplement will generally allow the material to go back into solution.

Legal Information

SER-TAIN is a trademark of Sigma-Aldrich Co. LLC
SERUM PLUS is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Proteasome inhibition for treatment of leishmaniasis, Chagas disease and sleeping sickness.
Khare S et al.
Nature, 537, 229-229 (2016)
Utilizing Chemical Genomics to Identify Cytochrome b as a Novel Drug Target for Chagas Disease.
Khare S et al.
PLoS Pathogens, 11, e1005058-e1005058 (2015)
Joseph Verdi et al.
Cell host & microbe, 28(1), 79-88 (2020-05-18)
Trypanosomiasis is a devastating neglected tropical disease affecting livestock and humans. Humans are susceptible to two Trypanosoma brucei subspecies but protected from other trypanosomes by circulating high-density lipoprotein (HDL) complexes called trypanosome lytic factors (TLFs) 1 and 2. TLFs contain
Nidhi Gupta et al.
Journal of the American Society of Nephrology : JASN, 31(9), 2065-2082 (2020-08-09)
Circulating APOL1 lyses trypanosomes, protecting against human sleeping sickness. Two common African gene variants of APOL1, G1 and G2, protect against infection by species of trypanosomes that resist wild-type APOL1. At the same time, the protection predisposes humans to CKD

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service