Skip to Content
Merck
All Photos(1)

Key Documents

32201

Sigma-Aldrich

Acetone

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH3
CAS Number:
Molecular Weight:
58.08
Beilstein:
635680
EC Number:
MDL number:
UNSPSC Code:
12352115
eCl@ss:
39021201
PubChem Substance ID:
NACRES:
NA.02
Assay:
≥99.5% (GC)
technique(s):
UV/Vis spectroscopy: suitable
bp:
56 °C/760 mmHg (lit.)
vapor pressure:
184 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

ACS reagent
puriss. p.a.

Quality Level

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

vapor density

2 (vs air)

vapor pressure

184 mmHg ( 20 °C)

Assay

≥99.5% (GC)

form

liquid

expl. lim.

13.2 %

technique(s)

UV/Vis spectroscopy: suitable

impurities

≤0.0002% KMnO4 red. matter (as O)
≤0.0002% benzene
≤0.0008% free alkali (as NH3)
≤0.001% aldehydes (as HCHO)
≤0.001% non-volatile matter
≤0.0015% free acid (as CH3COOH)
≤0.01% ethanol (GC)
≤0.05% 2-propanol
≤0.05% methanol
≤0.2% water (Karl Fischer)

refractive index

n20/D 1.3580-1.3600
n20/D 1.359 (lit.)

pH

5-6 (20 °C, 395 g/L)

bp

56 °C/760 mmHg (lit.)

mp

−94 °C (lit.)

density

0.791 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 ppm
B: ≤0.02 ppm
Ba: ≤0.1 ppm
Bi: ≤0.1 ppm
Ca: ≤0.5 ppm
Cd: ≤0.05 ppm
Co: ≤0.01 ppm
Cr: ≤0.02 ppm
Cu: ≤0.02 ppm
Fe: ≤0.1 ppm
K: ≤0.5 ppm
Li: ≤0.1 ppm
Mg: ≤0.1 ppm
Mn: ≤0.01 ppm
Mo: ≤0.1 ppm
Na: ≤0.5 ppm
Ni: ≤0.02 ppm
Pb: ≤0.02 ppm
Sn: ≤0.1 ppm
Sr: ≤0.1 ppm
Zn: ≤0.05 ppm

SMILES string

CC(C)=O

format

neat

InChI

1S/C3H6O/c1-3(2)4/h1-2H3

InChI key

CSCPPACGZOOCGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Mixtures of TiO2 (P25) with rare earth oxides (La2O3 or Y2O3) calcined at 700°C or 650°C have been reported to efficiently photocatalyze oxidation of acetone.[1] Its aldol condensation in vapor-phase has been investigated over MgO promoted with alkali (Li, Na, K and Cs) or alkaline earth (Ca, Sr and Ba) metal ions.[2] It undergoes Aldol condensation in the presence of Mg-Al layered double hydroxides (LDH) as catalysts and Cl- and/or CO32- as compensating anions to afford diacetone alcohol and mesityl oxide.[3] Enantioselective Aldol condensation of acetone with various isatins in the presence of dipeptides (catalyst) has been described.[4]

Application

Acetone was used in the synthesis of Ga (Gallium)-DOTATATE (where DOTA=1,4,7,10-tetraazacyclo- dodecane -1,4,7,10-tetraacetic acid) chemicals.[5] It may be used in an assay for determination of ester groups in lipids by spectrophotometric methods.[6]
Acetone′s luminesence intensity is dependent upon the solution components . The absorption of UV light by acetone, results in its photolysis and the production of radials .

Other Notes

The article number 32201-4X2.5L will be discontinued. Please order the single bottle 32201-2.5L which is physically identical with the same exact specifications.
The article number 32201-6X1L will be discontinued. Please order the single bottle 32201-1L which is physically identical with the same exact specifications.
The article number 32201-6X1L-M will be discontinued. Please order the single bottle 32201-1L-M which is physically identical with the same exact specifications.

greener alternative product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Gallium-68 DOTATATE Production with Automated PET Radiopharmaceutical Synthesis System: A Three Year Experience.
Aslani A, et al.
Asia Oceania Journal of Nuclear Medicine and Biology, 2(2), 75-86 (2014)
Base catalysis for the synthesis of α,β-unsaturated ketones from the vapor-phase aldol condensation of acetone.
Di Cosimo JI, et al.
Applied Catalysis A: General, 137(1), 149-166 (1996)
A simplified spectrophotometric determination of ester groups in lipids.
F SNYDER et al.
Biochimica et biophysica acta, 34, 244-245 (1959-07-01)
Gianluigi Luppi et al.
The Journal of organic chemistry, 70(18), 7418-7421 (2005-08-27)
[reaction: see text] The aldol condensation of acetone with several isatins is described. The desired compound was obtained in quantitative yield and with good enantioselectivities up to 77%. The best results were obtained with 10 mol % H-D-Pro-L-beta3-hPhg-OBn as a
Aldol condensation of acetone over layered double hydroxides of the meixnerite type.
Tichit D, et al.
Applied Clay Science, 13(5), 401-415 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service