Skip to Content
Merck
All Photos(1)

Key Documents

33211

Sigma-Aldrich

Ethyl acetate

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)

Synonym(s):

EtOAc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOC2H5
CAS Number:
Molecular Weight:
88.11
Beilstein:
506104
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.5% (GC)
bp:
76.5-77.5 °C (lit.)
vapor pressure:
73 mmHg ( 20 °C)

grade

ACS reagent
puriss. p.a.

Quality Level

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

vapor density

3 (20 °C, vs air)

vapor pressure

73 mmHg ( 20 °C)

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

801 °F

expl. lim.

2.2-11.5 %, 38 °F

impurities

≤0.001% non-volatile matter
≤0.0045% free acid (as CH3COOH)
≤0.05% water (Karl Fischer)
≤0.1% ethanol (GC)
≤0.1% isobutylalcohol
≤0.1% methanol
≤0.1% methyl acetate

color

APHA: >10

refractive index

n20/D 1.3710-1.3730
n20/D 1.3720 (lit.)

bp

76.5-77.5 °C (lit.)

mp

−84 °C (lit.)

density

0.902 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
B: ≤0.02 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sn: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

CCOC(C)=O

suitability

passes test for reaction against H2SO4

format

neat

InChI

1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3

InChI key

XEKOWRVHYACXOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Steroid Components of Marine-Derived Fungal Strain Penicillium levitum N33.2 and Their Biological Activities.: This article discusses the isolation and identification of steroid components using ethyl acetate, showcasing the role of analytical chemistry in understanding bioactive compounds from marine fungi, crucial for developing analytical standards and protocols (Hoang et al., 2023).

  • Chemical Defense against Herbivory in the Brown Marine Macroalga Padina gymnospora Could Be Attributed to a New Hydrocarbon Compound.: This research uses ethyl acetate in the extraction of novel hydrocarbon compounds, providing insights into chemical defenses of marine algae, relevant in environmental analytical chemistry for natural product isolation and analysis (Pereira et al., 2023).

  • Isolation and Characterization of One New Natural Compound with Other Potential Bioactive Secondary Metabolites from Glycosmis cyanocarpa (Blume) Spreng. (Family: Rutaceae).: This paper focuses on the isolation of bioactive compounds using ethyl acetate, emphasizing its critical role in the standardization of extraction and characterization processes in phytochemical analysis (Islam et al., 2023).

Other Notes

The article number 33211-4X2.5L-M will be discontinued. Please order the single bottle 33211-2.5L-M which is physically identical with the same exact specifications.
The article number 33211-4X2.5L-R will be discontinued. Please order the single bottle 33211-2.5L-R which is physically identical with the same exact specifications.
The article number 33211-6X1L-M will be discontinued. Please order the single bottle 33211-1L-M which is physically identical with the same exact specifications.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Flavio G Rocha et al.
Biomaterials, 29(19), 2884-2890 (2008-04-09)
Our group has previously created a functional neointestine that is capable of restoring absorptive function. However, the endogenous level of vascular endothelial growth factor (VEGF) is markedly reduced in the construct compared to native bowel. Therefore, we wanted to locally
Tsukasa Horiyama et al.
PloS one, 9(9), e108642-e108642 (2014-09-27)
Escherichia coli produces the iron-chelating compound enterobactin to enable growth under iron-limiting conditions. After biosynthesis, enterobactin is released from the cell. However, the enterobactin export system is not fully understood. Previous studies have suggested that the outer membrane channel TolC
K J S Anand et al.
British journal of anaesthesia, 101(5), 680-689 (2008-08-30)
Relationships between plasma morphine concentrations and neonatal responses to endotracheal tube (ETT) suctioning are unknown in preterm neonates. Ventilated preterm neonates (n=898) from 16 centres were randomly assigned to placebo (n=449) or morphine (n=449). After an i.v. loading dose (100
Yuusuke Fujimoto et al.
PloS one, 7(11), e48685-e48685 (2012-11-13)
Combination therapy with ribavirin, interferon, and viral protease inhibitors could be expected to elicit a high level of sustained virologic response in patients infected with hepatitis C virus (HCV). However, several severe side effects of this combination therapy have been
Maarit Neuvonen et al.
PloS one, 9(8), e103743-e103743 (2014-08-27)
Bacterial cholesterol oxidase is commonly used as an experimental tool to reduce cellular cholesterol content. That the treatment also generates the poorly degradable metabolite 4-cholesten-3-one (cholestenone) has received less attention. Here, we investigated the membrane partitioning of cholestenone using simulations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service