Skip to Content
Merck
All Photos(5)

Key Documents

904082

Sigma-Aldrich

Acetone

greener alternative

BioRenewable, ACS reagent, ≥99.5%

Synonym(s):

Dimethyl ketone, Propanone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH3
CAS Number:
Molecular Weight:
58.08
Beilstein:
635680
MDL number:
UNSPSC Code:
12352115
NACRES:
NA.02
grade:
ACS reagent
Assay:
≥99.5%
bp:
56 °C/760 mmHg (lit.)
vapor pressure:
184 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

ACS reagent

Agency

suitable for EPA 1613

vapor density

2 (vs air)

vapor pressure

184 mmHg ( 20 °C)

Assay

≥99.5%

form

liquid

expl. lim.

13.2 %

greener alternative product characteristics

Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

renewable carbon content

100%

refractive index

n20/D 1.359 (lit.)

bp

56 °C/760 mmHg (lit.)

mp

−94 °C (lit.)

density

0.791 g/mL at 25 °C (lit.)

format

neat

greener alternative category

SMILES string

CC(C)=O

InChI

1S/C3H6O/c1-3(2)4/h1-2H3

InChI key

CSCPPACGZOOCGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is 100% biorenewable and thus aligns with "Safer Solvents and Auxiliaries" and "Use of Renewable Feedstocks". Click here for more information.
This ACS grade Acetone is a drop-in replacement for any Acetone application. Made from 100% renewable carbon (verified through C14 ASTM D6866-16 testing) and produced in an environmentally friendly way that reduces the carbon footprint of production and reliance on fossil-fuels.
Because it is not from petroleum, it is free from many non-renewable contaminants, such as benzene, phthalates and other phenols, leading to higher quality product.

Application

  • Used as a solvent and/or building block in organic syntheses, such as SN2 reactions and Jones oxidation
  • Mobile phase in Thin layer chromatography (TLC) and other analytical applications



This is a plant waste-derived acetone, that acts as a drop-in replacement for any acetone application with no change in procedure or equipment necessary.
Acetone′s luminesence intensity is dependent upon the solution components . The absorption of UV light by acetone, results in its photolysis and the production of radials .

Features and Benefits

  • Made from Renewable Resource
  • Replacement for Petroleum-based Acetone
  • Renewable Carbon Content - ≥99.9%
  • ASTM D6866 - Standard Test Methods for Determining the Biobased Content

related product

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

1–4 of 4 Questions  
  1. what type of packaging or container is used for the acetone BioRenewable, ACS reagent, with a purity of ≥99.5% (SKU 904082-4L)?

    1 answer
    1. The container for 904082-4L is a 4 L Narrow Mouth Amber Glass bottle with a Black PP CAP and PTFE LINER.

      Helpful?

  2. What is a BioRenewable solvent?

    1 answer
    1. BioRenewable solvents are sourced from renewable and sustainable biobased materials, such as soy/palm oil, wood waste, hay or corn byproducts, etc. It is important to highlight that our BioRenewable products do not compete with food production.

      Helpful?

  3. Do BioRenewable solvents have a similar quality to petroleum-based solvents?

    1 answer
    1. BioRenewable solvents can have the same or higher purity level compared to traditional solvents. These solvents are free of the many byproducts of petroleum manufacturing, such as benzene, aldehydes, and ethers.

      Helpful?

  4. Why should you use BioRenewable solvents?

    1 answer
    1. You will contribute to create a sustainable world, significantly lowering environmental impact. Biorenewable solvents also decrease petroleum sources dependency, have higher recovery rates, and/or are safer for handling and disposal.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service