Skip to Content
Merck
Acetone puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)

Acetone

Acetone puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Linear Formula:
CH3COCH3
CAS Number:
Molecular Weight:
58.08
EC Number:

Products

Product Category
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-20 of 28
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2