216674
Silver acetate
ReagentPlus®, 99%
Synonym(s):
Acetic acid silver salt
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
CH3COOAg
CAS Number:
Molecular Weight:
166.91
Beilstein:
3595636
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.21
Assay:
99%
form:
powder or chunks
Recommended Products
Quality Level
product line
ReagentPlus®
Assay
99%
form
powder or chunks
reaction suitability
reagent type: catalyst
core: silver
SMILES string
CC(O[Ag])=O
InChI
1S/C2H4O2.Ag/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1
InChI key
CQLFBEKRDQMJLZ-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Used in a novel preparation of highly reflective, conductive silvered polymer films. Also effectively catalyzes cycloaddition reactions of isocyanoacetates with a variety of olefins.
Legal Information
ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Chemistry of Materials, FL501-FL501 (1999)
Silver acetate-assisted formation of amides from acyl chlorides.
Leggio A, et al.
Tetrahedron Letters, 56(1), 199-202 (2015)
Grigg, R. et al.
Tetrahedron, 55, 2025-2025 (1999)
Silver acetate catalysed cycloadditions of isocyanoacetates.
Grigg R, et al.
Tetrahedron, 55(7), 2025-2044 (1999)
Study of thermal decomposition of silver acetate.
Logvinenko V, et al.
Journal of Thermal Analysis and Calorimetry, 90(3), 813-816 (2007)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service