Skip to Content
Merck
All Photos(3)

Key Documents

209139

Sigma-Aldrich

Silver nitrate

ACS reagent, ≥99.0%

Synonym(s):

Nitric acid silver(I) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AgNO3
CAS Number:
Molecular Weight:
169.87
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
form:
solid

grade

ACS reagent

vapor density

5.8 (vs air)

Assay

≥99.0%

form

solid

reaction suitability

reagent type: catalyst
core: silver

clarity of soln

passes test

impurities

Free acid, passes test
≤0.01% not HCl pptd.

mp

212 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤5 ppm
sulfate (SO42-): ≤0.002%

cation traces

Cu: ≤2 ppm
Fe: ≤2 ppm
Pb: ≤0.001%

SMILES string

[O-][N+]([O-])=O.[Ag+]

InChI

1S/Ag.NO3/c;2-1(3)4/q+1;-1

InChI key

SQGYOTSLMSWVJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silver nitrate, also known as Nitric acid silver(I) salt, is an odorless, colorless, or white versatile inorganic compound widely used as an oxidant and cauterizing agent due to its sclerosing, dehydrating, and escharotic properties. It is commonly used as a catalyst in the hydrolytic oxidation of organosilanes, leading to the production of high hydrogen yield and organosilanols.

Application

Silver nitrate can be used as a catalyst:      
  • For the Friedel-Crafts acylation of benzene derivatives to synthesize corresponding ketones.     
  • In the hydrolytic oxidation of organosilanes to produce hydrogen.

It can be used to prepare some supported acid catalysts for various organic transformations. For example:      
  • AgNO3–SiO2 based catalyst is used to dearomatize spirocyclization.      
  • Polyaniline-AgNO3-PTSA (para-toluene sulfonic acid) catalyst is used to synthesize anti-β-amino ketones via the three-component Mannich reaction of aromatic aldehydes, aromatic amines, and cyclohexanone.

AgNO3 is also used as a precursor salt to synthesize silver nanoparticles.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Ox. Sol. 2 - Repr. 1B - Skin Corr. 1A

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Convenient Method for the Friedel-Crafts Acylation of Benzene Derivatives Using Silver Nitrate as Catalyst
Rai KML, et al.
Synthetic Communications, 41(7), 953-955 (2011)
Catalytic hydrogen evolution from hydrolytic oxidation of organosilanes with silver nitrate catalyst
Teo AKL and Fan WY
Royal Society of Chemistry Advances, 4(71), 37645-37648 (2014)
Synthesis and electrical properties of uniform silver nanoparticles for electronic applications
Chen D, et al.
J. Mater. Sci., 44(4), 1076-1081 (2009)
Catalytic hydrogen evolution from hydrolytic oxidation of organosilanes with silver nitrate catalyst
Alan Kay Liang T, et al.
Royal Society of Chemistry Advances, 4, 37645-37648 (2014)
A Novel Polyaniline-Silver Nitrate-p-Toluenesulfonic Acid Salt as Recyclable Catalyst in the Stereoselective Synthesis of ?-Amino Ketones:?One-Pot? Synthesis in Water Medium
Palaniappan S and Rajender B
Advanced Synthesis & Catalysis, 352(14-15), 2507-2514 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service