Skip to Content
Merck
All Photos(1)

Key Documents

12249

Sigma-Aldrich

Potassium chromate

puriss., ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2CrO4
CAS Number:
Molecular Weight:
194.19
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.55
Assay:
≥99%
form:
powder or crystals

vapor density

6.7 (vs air)

Quality Level

grade

puriss.

Assay

≥99%

form

powder or crystals

loss

≤0.5% loss on drying, 105 °C

pH

8.5-10.0 (20 °C, 5%)

mp

971 °C (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤200 mg/kg

cation traces

Ca: ≤50 mg/kg

SMILES string

[K+].[K+].[O-][Cr]([O-])(=O)=O

InChI

1S/Cr.2K.4O/q;2*+1;;;2*-1

InChI key

XMXNVYPJWBTAHN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Analysis of the Effects of Cr(VI) Exposure on mRNA Modifications.: This research investigates the impact of potassium chromate exposure on mRNA modifications using sophisticated analytical chemistry techniques, revealing insights into the molecular mechanisms of chromium toxicity and its implications for human health (Chen et al., 2019).
  • Determination of urine oxalate level in rats with renal calcium oxalate calculus by high-performance liquid chromatography.: Utilizing potassium chromate in the chromatographic analysis, this study presents a method for accurately measuring urine oxalate levels, crucial for research on kidney stone prevention (Cao et al., 2015).
  • Shotgun proteomic analysis unveils survival and detoxification strategies by Caulobacter crescentus during exposure to uranium, chromium, and cadmium.: Incorporates potassium chromate to study microbial resistance mechanisms through proteomic profiling, providing valuable data for bioremediation strategies (Yung et al., 2014).
  • Analytical study of modern paint layers on metal knight shields: the use and effect of titanium white.: Features methods where potassium chromate aids in the identification of chemical components in cultural heritage conservation studies (Lauridsen et al., 2014).

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Irrit. 2 - Muta. 1B - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Redetermination of the structures of potassium sulphate and potassium chromate: the effect of electrostatic crystal forces upon observed bond lengths.
McGinnety JA.
Acta Crystallographica Section B, Structural Science, Crystal Engineering and Materials, 28(9), 2845-2852 (1972)
Comparative Study of Factors Influencing the Action of Corrosion Inhmitors for Mild Steel in Neutral Solution: II. Potassium Chromate.
Mercer AD and Jenkins IR.
British Corrosion Journal, 3(3), 130-135 (1968)
Eagleson M.
Concise Encyclopedia Chemistry, 881-881 (1994)
A new method of potassium chromate production from chromite and KOH-KNO3-H2O binary submolten salt system.
Sun Z, et al.
AIChE Journal, 55(10), 2646-2656 (2009)
Xiao-Zhang Yu et al.
Ecotoxicology (London, England), 19(1), 61-68 (2009-09-04)
The removal of hexavalent and trivalent chromium from hydroponic solution by plants to changes in temperature was investigated. Pre-rooted hybrid willows (Salix matsudana Koidz x alba L.) were exposed to a nutrient solution spiked with potassium chromate (K(2)CrO(4)) or chromium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service